Interested in a bulk or custom version of this product? Request pricing.

ARSA, Recombinant Mouse Protein, His tag

Catalog number: 50018M08H50

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Arylsulfatase A (ARSA), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Ser 506) of mouse ARSA precursor (NP_033843.2) fused to the polyhistidine tag at the C-terminus.

Activity: Measured by its ability to cleave p-Nitrocatechol Sulfate (PNCS)
. The specific activity is >100 pmoles/min/µg
.

Formulation: Lyophilized in 25 mM Tris, 150 mM NaCl, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.

This protein is manufactured by Sino Biological Inc.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Endotoxin Level: < 1 EU/µg
Expression: Mammalian
Form: Lyophilized
Gene Aliases: Arsa, AS-A, ASA, AW212749, As-2, As2, C230037L18Rik, TISP73
Gene Symbol: ARSA
Molecular Weight: 53.5 kDa
Product Line: Sino Biological®
Product Size: 50 µg
Protein Family: Other Proteins
Protein Name: Arylsulfatase A
Protein Subtype: Other Proteins
Protein Tag or Fusion: His Tag
Purity: ≥ 90% by SDS-PAGE
Research Category: Neurobiology, Other Research
Species: Mouse
Validated Application: Assay Standard

Contents & storage

Includes 1 vial of lyophilized protein. Store at -20 °C.

Documents

Manuals & protocols