M104 Muffle Furnaces
Thermo Scientific™  Related applications: Lab Ovens Furnaces
Protect against sample contamination with Thermo Scientific™ M104 Muffle Furnaces, an abrasion-resistant ceramic annealing chamber. Designed for minimum space requirements, they are ideal for chemical analysis, annealing loss determination, materials testing and ashing processes. Standard models are equipped with electronic controller, digital display and upper limit cut-out; deluxe models are available with electronic or programmable controller, timer and exhaust fan.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Introducing shared lists
Back to top
Description
Key Features:
  • Environmentally friendly and economical—asbestos-free insulation minimizes energy consumption and keeps exterior temperature low
  • Easy handling and loading via tilting door that also protects users from hot inside surface
  • Reaches rated temperature of 1000°C in 110 min.
  • Working volume of 3.5L
  • Standard model equipped with electronic controller, digital display and upper limit cut-out; deluxe models available with electronic or programmable controller, timer, and exhaust fan
  • M104G models feature glazed annealing muffle for use in analytical processes requiring extreme purity
  • Can be set up under a laboratory fume hood.
Recommended for:
  • Chemical analysis
  • Annealing loss determination
  • Materials testing Ashing processes
Ordering Information:
Product is not available in North America.
Documents
Manuals & protocols