Fermacidal Disinfectant for all CO<sub>2</sub> Incubators, 500 mL
Thermo Scientific™

Fermacidal Disinfectant for all CO2 Incubators, 500 mL

Quaternary ammonium disenfectant for all Thermo Scientific™ CO2 Incubators
Have Questions?
Número do catálogoCapacity (Metric)
50051939500 mL
Número do catálogo 50051939
Preço (BRL)
-
Capacity (Metric):
500 mL
Ready-to-use, quaternary ammonium 500 mL disinfectant agent for use in all Thermo Scientific™ CO2 Incubators.
  • Quaternary ammonium disinfectant
  • For use in all Thermo Scientific™ CO2 Incubators
  • Spray in, swab off, or rinse through
Especificações
Capacity (English)0.585 kg
DescriptionReady-to-use, 500 mL disenfectant agent
For Use With (Equipment)All Thermo Scientific™ CO2 Incubators
Capacity (Metric)500 mL
Unit SizeEach