Interested in a bulk or custom version of this product? Request pricing.

CXCL2 Recombinant Mouse Protein, N-His.SUMO Tag

Catalog number: 50070M28E25

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Chemokine (C-X-C Motif) Ligand 2 (CXCL2), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

This recombinant protein was expressed from a DNA sequence encoding the mature form of mouse CXCL2 (P10889-1) (Ala 28-Asn 100) fused to a N-terminal polyhistide-tagged SUMO tag at the N-terminus.

Activity: The activity of this protein has not been tested.

Formulation: Lyophilized in 0.1% TFA, pH 2.9, 5% Mannitol, 5% Trehalose, 0.02% Tween 80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

This protein is manufactured by Sino Biological Inc.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Research Category: Cardiovascular Research, Immunology, Inflammation, Neurobiology, Oncology
Endotoxin Level: < 1 EU/µg
Expression: E. coli
Form: Lyophilized
Gene Aliases: CINC-2a, GROb, Gro2, MIP-2, MIP-2a, Mgsa-b, Mip2, Scyb, Scyb2
Gene ID: 20310
Gene Symbol: CXCL2
Molecular Weight: 21.2 kDa
Product Line: Sino Biological®
Product Size: 5 x 5 µg
Protein Family: Chemokines & Receptors
Protein Form: Full Length, Recombinant
Protein Name: Chemokine (C-X-C motif) ligand 2
Protein Subtype: GRO⁄KC⁄CINC Chemokines, NFκB Pathway Proteins
Protein Tag or Fusion: His Tag, SUMO Tag
Purification: PAGE
Purity: ≥ 90% by SDS-PAGE
Species: Mouse
Tag Position: N-Terminal
Validated Application: Assay Standard

Contents & storage

Includes 5 vials of protein in suspension buffer. Store at -20 °C..

Documents

Manuals & protocols