Herasafe™ KS Options and Accessories
Actual product may vary
Herasafe™ KS Options and Accessories
Herasafe™ KS Options and Accessories
Herasafe™ KS Options and Accessories
Herasafe™ KS Options and Accessories
Thermo Scientific™
Herasafe™ KS Options and Accessories
Worksurfaces, exhaust transitions and baskets
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct Type
50076398900 mm Flat Worksurface Module
50073757300 mm Vubration Damped Worksurface
50073667600 mm Flat Worksurface Module
50073666150 mm Flat Worksurface Module
50073669900 mm Indented Worksurface Module
50078795KS18 Thimble Duct Exhaust Acc.
50078900Utility Basket
500736701200 mm Indented Worksurface Module
50076162600 mm Vubration Damped Worksurface
50073770KS12/KS15 Thimble Duct Exhaust Acc.
191780Exhaust Alarm for Thimble Duct
50073668600 mm Indented Worksurface Module
50078796KS9 Thimble Duct Exhaust Acc.
500736711500 mm Indented Worksurface Module
50078901Utility Basket with Hanging Bar
500787501800 mm Indented Worksurface Module
Catalog number 50076398
Price (MXN)
-
Product Type:
900 mm Flat Worksurface Module
Worksurfaces, exhaust transitions and baskets
Specifications
DescriptionFlat Worksurface Module 900 mm Wide (316SS)
Housing Material316 SS
Width (Metric)900 mm
Width (Metric) Exterior900 mm
Product Type900 mm Flat Worksurface Module
Unit SizeEach