Additional Activated Carbon Filter
Additional Activated Carbon Filter
Additional Activated Carbon Filter
Thermo Scientific™
Additional Activated Carbon Filter
Additional exhaust carbon filters can be fitted on top of the BSC, but should only be used for traces of solvents being used as an adjunct to microbiological study.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)
500787830.9 m Herasafe KS/Safe 2020 Cabinets
50073773Retrofit to 1.2/1.5 m Cabinets
Catalog number 50078783
Price (MXN)
-
For Use With (Equipment):
0.9 m Herasafe KS/Safe 2020 Cabinets
Additional exhaust carbon filters can be fitted on top of the BSC, but should only be used for traces of solvents being used as an adjunct to microbiological study.
  • Additional Activated Carbon Filter
  • Fittings Included.
  • Additional exhaust carbon filters can be fitted on top of the BSC, but should only be used for traces of solvents being used as an adjunct to microbiological study.
Specifications
Filter TypeCarbon
For Use With (Equipment)0.9 m Herasafe KS/Safe 2020 Cabinets
IncludesFittings
Product TypeFilter
Certifications/ComplianceEN 12469
Unit SizeEach