Thermo Scientific™
Cytomat™ 6004 DR Automated Incubator
 Related applications:
Lab Automation
The Thermo Scientific™ Cytomat™ C600x series automated incubator is a mid size system with a capacity for 189 standard microplates. This series offers several models with various temperature ranges and gate positions.
The standard models are bench top and space-saving under bench solutions are also available. Its design and unsurpassed temperature uniformity and stability ensure that your processes can be fully reproduced, providing you with the same high quality results every time.
This series established the Cytomat family as the leader in wide range automated incubators, proven to provide extremely uniform and stable temperature, humidity and CO2 conditions in hundreds of automated systems worldwide.
The Cytomat 6004 DR automated incubator provides an ideal stable constant humidity level of 6% r.H. This extremely dry environment minimizes absorption of water by hydrophilic compounds (e.g. DMSO), thereby reducing assay variance.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
Matches your Capacity with 9 Stacker
  • 252pcs. 1536-well plates
  • 189pcs. 96-well or 384-well plates
  • 135pcs. transwell or insert plates
  • 90pcs. deep-well
Further stacker types on request
Mix and match different pitch stackers
Stable low rel. humidity conditions at ambient temperature
  • maintains constant humidity level of 6% r.H. at ambient temperature to minimize absorption of water by hydrophilic compounds (e.g. DMSO)
Access Time
  • Below 20sec.
Contamination Prevention
  • Easy to clean - smooth stainless steel surface
  • Solid copper inner chamber (option)
  • H2O2-flange (Option)
Flexible Stacker Design
  • Compatible with existing Cytomat stacker
  • Standard or customized stacker pitch sizes
  • Mix and match different pitch stackers
  • Autoclaveable, stainless steel
  • Allows for accurate temperature distribution with ventilation slots
Convenient
  • Internal and direct barcode reader can scan barcodes at any plate position inside the incubator the incubator, minimizing plate movement and provides superior sample and plate tracking
Easy to integrate
  • Intuitive software commands
  • SILA (Standard in Lab Automation) Compliant
  • Direct connection to a clean bench
  • Multiple transfer stations
Many Further Options
  • Many gate positions
  • SiLA interface option for fast and easy integration
  • Standard transfer station - delivers plates in portrait position
  • 90° (for landscape format) and 180" swap station
  • Linear extension with multiple length
Certification
  • CE
  • RoHS
  • Manufactured in ISO 9001 and ISO 13485 accredited facility
Documents
Manuals & protocols