Matrix™ ControlMate™ OLE Software Module
Thermo Scientific™
Matrix™ ControlMate™ OLE Software Module
Control all instrument parameters with simply with the drag-and-drop functionality of the Thermo Scientific™ Matrix™ ControlMate™ OLE Software Module.
Have Questions?
Catalog NumberDescription
50130023Matrix ControlMate OLE Software Module
Catalog number 50130023
Price (USD)
-
Description:
Matrix ControlMate OLE Software Module
Control all instrument parameters with simple commands in a tree structure with the simple drag-and-drop functionality of the Thermo Scientific™ Matrix™ ControlMate™ OLE Software Module. ControlMate commands are universal across all Thermo Scientific Matrix automation platforms, so when your application requires a new instrument, you will not waste time learning an entirely new operating system.
Benefits:
  • Simple commands in a tree structure
  • Utilizes simple drag-and-drop icons
  • Includes CD-ROM, security key and manual
Allows control of the following parameters:
  • Speed control of X, Y and Z plate movements
  • Control of piston speed
  • Tip position and tip touch-off in any microplate well
  • Aspiration/dispense volumes
  • Dwell time before and after aspiration or dispense
  • Repeat (incremental) or neat dispensing
  • Specific volume calibration for non-aqueous-based liquids
  • Quadrant offsets for 384, 864 and 1536 well plates
  • Mixing
  • Direction and speed control of peristaltic pumps
  • Edit and run lock password protection
  • Automatic shut down of all systems at the end of each run or after a system failure
  • Standard shared plate library that is easily modified
  • Run time clock that tracks and documents total protocol elapsed time
Includes:
CD-ROM, security key, and manual
Specifications
DescriptionMatrix ControlMate OLE Software Module
IncludesCD-ROM, security key, and manual
Product TypeOLE Software Module
Unit SizeEach