Search
Suitable for the most demanding laboratory applications, the Barnstead™ GenPure™ Pro water purification system delivers ultrapure 18.2 megohm-cm water with consistent quality.
Suitable for the most demanding laboratory applications, the Thermo Scientific™ Barnstead GenPure Pro water purification system delivers ultrapure 18.2 megohm-cm water with consistent quality. This ultrapure water system will deliver water from a small footprint. The GenPure Pro system can be used on a laboratory bench or on a wall. The new flexible dispenser offers maximum freedom while dispensing ultrapure water up to 24 in. (60cm) away from the unit.