Stainless Steel Reinforced Shelf
Thermo Scientific™
Stainless Steel Reinforced Shelf
Stainless Steel Reinforced Shelf
Have Questions?
Catalog NumberCapacity (Metric)
5015064314 kg
Catalog number 50150643
Price (MXN)
-
Capacity (Metric):
14 kg
Stainless Steel Reinforced Shelf
  • Stainless Steel Reinforced Shelf
  • For Heracell VIOS 250/240i CO2 Incubator
Specifications
For Use With (Equipment)Heracell VIOS 250/240 i CO2 Incubator
LightingNo Lighting
Oxygen ControlNone
TypeReinforced Shelf
CO2 Sensor TechnologyNo Sensor
Capacity (Metric)14 kg
MaterialStainless Steel
Relative HumidityNone
Unit SizeEach