Barnstead™ Pretreatment for Point of Use Water Purification Systems
Barnstead™ Pretreatment for Point of Use Water Purification Systems
Barnstead™ Pretreatment for Point of Use Water Purification Systems
Barnstead™ Pretreatment for Point of Use Water Purification Systems
Thermo Scientific™
Barnstead™ Pretreatment for Point of Use Water Purification Systems
Barnstead Pretreatment for Point of Use Water Purification Systems
Have Questions?
카탈로그 번호카트리지 유형
50157886Pretreatment Cartridge
카탈로그 번호 50157886
제품 가격(KRW)
2,130,000
온라인 행사
Ends: 31-Dec-2026
2,505,000
할인액 375,000 (15%)
Each
카트리지 유형:
Pretreatment Cartridge
제품 가격(KRW)
2,130,000
온라인 행사
Ends: 31-Dec-2026
2,505,000
할인액 375,000 (15%)
Each
Barnstead Pretreatment contains PreFilter, Hardness Stabilizer and Carbon Filter. The unit contains pressure regulator and pressure gauges. Space saving design comes with Wall Mount capability.
  • Use pretreatment when feed water quality doesn't match the requirements for your system
  • Contains pre-filter, hardness stabilizer and carbon filter
사양
카트리지 유형Pretreatment Cartridge
설명Pretreatment includes Particle Filter, Carbon Filter and Hardness Stabilizer Catridges ( D502113, D502114 and D502115 )
치수(길이 x 폭 x 높이)14.5 in x 13.3 in x 20.0 in.
용도(장비)Smart2Pure Pro
포함Pressure Regulator, Pressure Gauges, Three Cartridges and Wall Mount
제품 유형Water Purification System
중량(영국식 단위)24.5 lb.
중량(미터법)11.4 kg
Unit SizeEach