Interested in a bulk or custom version of this product? Request pricing.

FIGF Recombinant Mouse Protein, hIgG1-Fc Tag

Catalog number: 50157M01H50

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

C-fos induced growth factor (vascular endothelial growth factor D) (FIGF/VEGFD), recombinant mouse protein is supplied as a lyophilized powder. In general, recombinant proteins are suitable for use in studying protein-protein interactions, as a protein standard, or in cell biology research applications.

This recombinant protein was expressed from a DNA sequence encoding the mature form of mouse FIGF (P97946) (Phe 98-Ser 206) fused to the Fc region of human IgG1 at the N-terminus.

Activity: Measured by its binding ability in a functional ELISA. 1. Immobilized human VEGFR3-His at 10 µg/ml(100 µl/well) can bind mouse Fc-VEGFD. The EC50 of mouse VEGFD-Fc is 49 ng/ml. 2. Immobilized mouse VEGFR3-His at 10 µg/ml(100 µl/well) can bind mouse Fc-VEGFD. The EC50 of mouse VEGFD-Fc is 44 ng/ml.

Formulation: Lyophilized in 140 mM NaCl, 2.7 mM KCl, 10 mM Na2HPO4, 1.8 mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.



This protein is manufactured by Sino Biological Inc.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Research Category: Development
Endotoxin Level: < 1 EU/µg
Expression: Mammalian
Form: Lyophilized
Gene Aliases: RP23-202H24.2, AI325264, VEGF-D, Vegfd
Gene Symbol: FIGF
Molecular Weight: 40.6 kDa
Product Line: Sino Biological®
Product Size: 50 µg
Protein Family: Growth Factors & Receptors
Protein Form: Full Length, Recombinant
Protein Name: c-fos induced growth factor
Protein Subtype: Other Growth Factors
Protein Tag or Fusion: IgG1 Fc Tag
Purity: ≥ 90% by SDS-PAGE
Species: Mouse
Validated Application: Assay Standard

Contents & storage

Includes 1 vial of lyophilized protein. Store at -20 °C.

Documents

Manuals & protocols

Recommended products