Search
| Katalognummer | Zur Verwendung mit (Anwendung) |
|---|---|
| 50168119 | OPC UA-Integration |
Enable Open Platform Communication Unified Architecture (OPC™ UA) integration of Heracell VIOS and Forma Steri-Cycle CO2 Incubators using this Thermo Scientific™ Gateway accessory kit. Benefit from the full potential of incorporating lab equipment into your new or existing DCS, including paperless record keeping, remote monitoring, cloud analytics and more.
Product Features
Thermo Scientific™ Gateway for OPC UA enablement of Heracell VIOS and Forma Steri-Cycle CO2 Incubators utilizes the OPC UA interoperability standard to enable a secure and reliable exchange of real-time and historical data between Distributed Control Systems (DCS) and enterprise users.
What is OPC UA?
Open Platform Communication Unified Architecture (OPC™ UA) is a platform-independent, open industry standard for connectivity that enables machine-to-machine communication and information integration between various devices, software systems, and applications.
Advantages to connecting
The gateway accessory kit comes complete with a gateway device, USB 2.0 cable USB-A to USB-B connector, and 100-240V power supply. One kit is needed per CO2 Incubator connection.