Interested in a bulk or custom version of this product? Request pricing.

ACVR2B Recombinant Mouse Protein (without catalytic activity domain), hIgG1-Fc His tag

Catalog number: 50173M03H50

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Activin Receptor IIB (ACVR2B), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

Since this product is a truncated protein and does not contain intracellular catalytic activity domain, it is not suitable for use in enzyme activity studies.

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Thr 134) of mouse ACVR2B (NP_031423.1) precursor fused to the Fc region of human IgG1 at the C-terminus.

Activity: 1. Measured by its ability to bind biotinylated Human INHBA-his (Cat:10429-H08H) in functional ELISA.
2. Measured by its ability to bind biotinylated mouse INHBA-his (Cat:50659-M08H) in functional ELISA.

Formulation: 140 mM NaCl, 2.7 mM KCl, 10 mM Na2HPO4, 1.8 mM KH2PO4, pH7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.



This protein is manufactured by Sino Biological Inc.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Protein Family: Growth Factors & Receptors
Research Category: Signal Transduction
Product Line: Sino Biological®
Product Size: 50 µg

Contents & storage

Includes 1 vial of protein in suspension buffer. Store at -20°C.

Documents

Manuals & protocols