Interested in a bulk or custom version of this product? Request pricing.

FGF1 (aFGF) Recombinant Mouse Protein

Catalog number: 50179MNAE250

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Fibroblast growth factor 1 (FGF1/aFGF), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in performing cell based assays. It can also be used as a protein standard or in other research applications.

This recombinant protein was expressed from a DNA sequence encoding the mature form (Phe 16-Asp 155) of mouse FGF1 (NP_034327.1).

Activity: Measured in a cell proliferation assay using BALB/c 3T3 mouse fibroblasts. The ED50 for this effect is typically 0.05-0.25 ng/mL in the presence of 10 µg/ml heparin.

Formulation: Lyophilized in 20 mM MES buffer, pH 6.0, 200 mM NaCl, 5% Mannitol, 5% Trehalose, and 0.02% Tween-80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 25 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.



This protein is manufactured by Sino Biological Inc.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Research Category: Cardiovascular Research, Oncology, Stem Cell Research
Expression: E. coli
Form: Lyophilized
Gene Aliases: Dffrx, Fam, Fgf-1, Fgfa, Fgf1
Gene ID: 14164
Gene Symbol: FGF1
Molecular Weight: 16 kDa
Product Line: Sino Biological®
Product Size: 5 x 50 µg
Protein Family: Growth Factors & Receptors
Protein Form: Recombinant
Protein Name: Fibroblast growth factor 1
Protein Subtype: FGFs (Fibroblast Growth Factors)
Protein Tag or Fusion: Untagged
Purity: > 98% by SDS-PAGE
Species: Mouse
Validated Application: Assay Standard

Contents & storage

Store at -20 to -70 °C

Documents

Manuals & protocols

Recommended products