Interested in a bulk or custom version of this product? Request pricing.

CD200R1 Recombinant Mouse Protein, His tag

Catalog number: 50209M08H5

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Cluster of Differentiation 200 receptor 1 (CD200R1), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, studying protein-protein interactions, and cell biology research applications. It can also be used as an immunogen, or as a protein standard.

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Pro 238) of mouse CD200R1 (NP_067300.1) fused to a C-terminal polyhistidine tag at the C-terminus.

Activity: Measured by its ability to bind mouse CD200 in a functional ELISA. Immobilized mouse CD200-His at 0.2 µg/well can bind mouse CD200R1-His with a linear range of 0.8-4.0 µg/ml.

Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.



This protein is manufactured by Sino Biological Inc.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Protein Family: CD Proteins
Research Category: Immunology
Endotoxin Level: < 1 EU/µg
Expression: Mammalian
Form: Lyophilized
Gene Aliases: CD200R, Mox2r, OX2R
Gene ID: 57781
Gene Symbol: CD200R1
Molecular Weight: 24.6 kDa
Product Line: Sino Biological®
Product Size: 5 µg
Protein Form: Cell Surface Protein, Recombinant, Truncated
Protein Name: CD200 receptor 1
Protein Subtype: Glycoproteins
Protein Tag or Fusion: His Tag
Purity: ≥ 85% by SDS-PAGE
Species: Mouse
Validated Application: Assay Standard
Shipping Condition: Room Temperature

Contents & storage

Store at -20 to -70 °C

Documents

Manuals & protocols