Interested in a bulk or custom version of this product? Request pricing.

PCSK9 Recombinant Mouse Protein, His Tag

Catalog number: 50251M08H50

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Proprotein Convertase Subtilisin/Kexin Type 9 (PCSK9), recombinant mouse protein is supplied in frozen format. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

This recombinant protein was expressed from a DNA sequence encoding the full length of mouse PCSK9 (NP_705793.1) precursor (Met 1-Gln 694) fused to a C-terminal polyhistidine tag.

Activity: Measured by its binding ability in a functional ELISA. Immobilized mouse PCSK9 at 10 µg/ml (100 µl/well) can bind biotinylated recombinant human LDLR. The EC50 of biotinylated human LDLR is 0.12 µg/ml.

Formulation: 15 mM Tris, 90 mM NaCl, 50% glycerol, pH 7.5.

Reconstitution: No reconstitution needed. Store at -80°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.

This protein is manufactured by Sino Biological Inc.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Research Category: Cardiovascular Research, Genetic Diseases
Endotoxin Level: < 1 EU/µg
Expression: Mammalian
Form: Frozen
Gene Aliases: RP23-159J7.1, AI415265, AI747682, FH3, HCHOLA3, MGC47409,Narc1
Gene Symbol: PCSK9
Molecular Weight: 72.6 kDa
Product Line: Sino Biological®
Product Size: 50 µg
Protein Name: proprotein convertase subtilisin/kexin type 9
Protein Subtype: Proteases
Protein Tag or Fusion: His Tag
Purity: ≥ 90% by SDS-PAGE
Species: Mouse
Validated Application: Assay Standard

Contents & storage

Store at -80 °C.

Documents

Manuals & protocols