Interested in a bulk or custom version of this product? Request pricing.

PVR (CD155) Recombinant Mouse Protein, hIgG1-Fc.His Tag

Catalog number: 50259M03H250

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top


Poliovirus receptor (PVR/CD155), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in studying protein-protein interactions, and cell biology research applications. It can also be used as a protein standard.

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Arg 345) of mouse PVR precursor (NP_081790.1) precursor fused to the polyhistidine-tagged Fc region of human IgG1 at the C-terminus.

Activity: Measured by its binding ability in a functional ELISA . Immobilized mouse DNAM1 at 2 µg/ml(100 µl/well) can bind mouse PVR with a linear range of 6.4-160 ng/ml.

Formulation: Lyophilized in 50 mM Tris, 10 mM NaCl, pH 7.5, 100 MM Glycine, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.

This protein is manufactured by Sino Biological Inc.

For Research Use Only. Not for use in diagnostic procedures.


Form: Lyophilized
Purity: ≥ 85% by SDS-PAGE
Gene ID: 52118
Species: Mouse
Expression: Mammalian
Gene Symbol: PVR
Gene Aliases: 3830421F03Rik, CD155, D7Ertd458e, HVED, PVS, Taa1, Tage4, mE4
Product Size: 5 x 50 µg
Protein Form: Truncated, Cell Surface Protein, Recombinant
Protein Family: CD Proteins
Endotoxin Level: < 1 EU/µg
Protein Subtype: Viral Host Cell Receptors
Molecular Weight: 63 kDa
Research Category: Immunology, Oncology, Virology, Microbiology, Neurobiology
Protein Tag or Fusion: His Tag, IgG1 Fc Tag
Validated Application: Assay Standard
Shipping Condition: Room Temperature

Contents & storage

Store at -20 to -70 °C


Manuals & protocols