Interested in a bulk or custom version of this product? Request pricing.

SCGB1A1 Recombinant Mouse Protein, His Tag

Catalog number: 50291M08H50

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Secretoglobin, Family 1A, Member 1 (Uteroglobin) (SCGB1A1), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications

This recombinant protein was expressed from a DNA sequence encoding the mouse SCGB1A1 (Q06318) (Met 1-Phe 96) fused to a polyhistidine tag at the C-terminus.

Activity: Measured by the ability of the immobilized protein to support the adhesion of the A549 human lung carcinoma cell line. When 5 x 104 cells/well are added to SCGB1A1 coated plates (5 µg/ml with 100 µl/well), approximately >40% will adhere after 1 hour at 37 °C.

Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

This protein is manufactured by Sino Biological Inc.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Endotoxin Level: < 1 EU/µg
Expression: Mammalian
Form: Lyophilized
Gene Aliases: CC10, CC16, CCSP, PCB-BP, UG, UGB, Utg
Gene ID: 22287
Gene Symbol: SCGB1A1
Molecular Weight: 9.8 kDa
Product Line: Sino Biological®
Product Size: 50 µg
Protein Family: Other Proteins
Protein Form: Full Length, Recombinant
Protein Name: secretoglobin, family 1A, member 1 (uteroglobin)
Protein Tag or Fusion: His Tag
Purification: PAGE
Purity: ≥ 85% by SDS-PAGE
Research Category: Immunology, Signal Transduction
Species: Mouse
Tag Position: C-Terminal
Validated Application: Assay Standard

Contents & storage

Includes 1 vials of protein in suspension buffer. Store at -20 °C..

Documents

Manuals & protocols