Regulatory requirements vary by country; products may not be available in your geographic area.

Abortion (BoHV1, BVDV, BoHV4) ELISA Kit

Catalog number: 5030450

 Related applications: Animal Health

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top


The Abortion (BoHV1, BVDV, BoHV4) ELISA Kit is an indirect ELISA kit for the serodiagnosis of BoHV1, BVDV, BoHV4 in cattle. The test uses 96-well microtiter plates sensitized by the three pathogens; 2 rows with BoHV-1, 2 with BVDV, 2 with BHV-4 and 2 with a negative control. The diluted test sera are incubated in the wells and antibodies present in the sample bind to the coated antigen. Bound antibodies are detected using a peroxidase-labeled monoclonal anti-bovine IgG1. Bound conjugate is visualized by adding chromogen (TMB) substrate. The intensity of the resulting blue color is proportionate to the titer of the specific antibody.
For Veterinary Use Only.


Species: Cattle⁄Bovine
Product Size: 48 samples
Protein Family: Other Proteins
Detection Method: Colorimetric


Manuals & protocols