Regulatory requirements vary by country; products may not be available in your geographic area.

Coronavirus Rapid Test Kit

Catalog number: 5030540

 Related applications: Animal Health

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Coronavirus Rapid Test Kit is a chromatographic lateral flow immunoassay is designed to detect Coronavirus in calf stool. The test uses two monoclonal antibodies, one as conjugate and one to capture the virus on the strip. This enables excellent specificity and reliable results. The test requires minimal hands-on-time and results are available typically in 10 minutes.
For Veterinary Use Only.

Specifications

Target Organism Class: Bovine Coronavirus
Detection Method: Colloidal Gold
Disease: Viral Infection
Product Line: Bio-X, Prionics™
Product Size: 10 strips
Species: Cattle⁄Bovine

Documents

Manuals & protocols