Regulatory requirements vary by country; products may not be available in your geographic area.

E. coli F5 (K99) Rapid Test Kit

Catalog number: 5030550

 Related applications: Animal Health

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The E. coli F5 (K99) Rapid Test Kit is a lateral immunochromatography test for the detection of E. coli F5 (K99) attachment factor. The test is designed to detect E. coli F5 attachment factor in calf, small ruminant and pig stool. The use of a monoclonal antibody as conjugate and to capture the bacterium on the strip enables excellent specificity and reliable results. The test requires minimal hands-on-time and results are available typically in 10 minutes.
For Veterinary Use Only.

Specifications

Target Organism Class: E. coli F5
Detection Method: Colloidal Gold
Disease: Bacterial Infection
Product Line: Bio-X, Prionics™
Product Size: 10 strips
Species: Cattle⁄Bovine, Swine⁄Porcine

Documents

Manuals & protocols