Regulatory requirements vary by country; products may not be available in your geographic area.

Bovine Parainfluenza 3 (PI3) ELISA Kit

Catalog number: 5030760

 Related applications: Animal Health

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Bovine Parainfluenza 3 (PI3) ELISA Kit is an ELISA kit for serodiagnosis of bovine Parainfluenza 3 (PI3) in cattle. This indirect test is used to follow seroconversions in paired sera or milk samples. The use of highly purified PI3 virus enables excellent specificity and reliable results. The test uses Protein G, which recognizes most immunoglobulin isotypes, for detection. The test is fast and easy to use with minimal hands-on-time.

Samples are incubated in microtiter plates coated with purified, inactivated PI3 virus. Bound antibodies are detected using peroxidase-labeled Protein G. Binding is visualized by a colorimetric reaction after the addition of the substrate.
For Veterinary Use Only.

Specifications

Protein Family: Other Proteins
Assay Sensitivity: See product documentation
Gene ID: Non-Gene
Gene Symbol: Non-Gene
Immunoassay Kit Format: Various Other
Product Line: Bio-X, Prionics™
Product Size: 96 samples
Target (Kit Named): Bovine Parainfluenza 3 (PI3)
Target Species (Named): Bovine
Target Species (Validated): Bovine

Documents

Manuals & protocols