Regulatory requirements vary by country; products may not be available in your geographic area.

Mycoplasma bovis ELISA Kit

Catalog number: 5030810

 Related applications: Animal Health

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Mycoplasma bovis ELISA Kit is an ELISA kit for the serodiagnosis of Mycoplasma bovis in cattle. The kit is designed to follow seroconversion on paired sera or to evaluate the serological status of serum or milk. This reliable test is easy to use and requires minimal hands-on time. Results are available typically in 140 minutes for single or batch testing.

The indirect ELISA uses microtiter plates coated with recombinant protein. Milk or serum samples are incubated in the wells. Anti- Mycoplasma bovis antibodies present in the sample bind to the recombinant protein and are detected with peroxidase-labeled Protein G. Bound conjugate is visualized by adding chromogen (TMB) substrate. When anti- Mycoplasma bovis antibodies are present in the sample, a color signal will appear.
For Veterinary Use Only.

Specifications

Protein Family: Other Proteins
Assay Sensitivity: See product documentation
Gene ID: Non-Gene
Gene Symbol: Non-Gene
Immunoassay Kit Format: Various Other
Product Line: Bio-X, Prionics™
Product Size: 240 samples
Target (Kit Named): Mycoplasma bovis
Target Species (Named): Bovine
Target Species (Validated): Bovine

Documents

Manuals & protocols