Regulatory requirements vary by country; products may not be available in your geographic area.

BoHV4 ELISA Kit

Catalog number: 5030820

 Related applications: Animal Health

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The BoHV4 ELISA Kit is designed to follow seroconversion on paired sera or to evaluate the serological status of serum or milk. The use of purified virus helps produce excellent specificity and reliable results. Protein G used as conjugate recognizes most immunoglobulin isotypes. The test requires minimal hands-on-time and results are available typically in 140 minutes for single or batch testing.

Samples are incubated in microtiter plates coated with purified, inactivated virus. Bound antibodies present in the sample are detected using peroxidase labeled Protein G. Binding is visualized by a colorimetric reaction after the addition of the substrate.
For Veterinary Use Only.

Specifications

Protein Family: Other Proteins
Assay Sensitivity: See product documentation
Gene ID: Non-Gene
Gene Symbol: Non-Gene
Immunoassay Kit Format: Various Other
Product Line: Bio-X, Prionics™
Product Size: 96 samples
Target (Kit Named): BoHV4
Target Species (Named): Bovine
Target Species (Validated): Bovine

Documents

Manuals & protocols