Regulatory requirements vary by country; products may not be available in your geographic area.

Bovine Haptoglobine Quantitative ELISA Kit

Catalog number: 5030880

 Related applications: Animal Health

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Bovine Haptoglobine Quantitative ELISA Kit is a quantitative direct sandwich ELISA for assaying the haptoglobin concentration in bovine blood serum and milk samples. The test uses 96-well microplates sensitized with a polyclonal antibody directed against bovine haptoglobin. This antibody enables specific capture of the protein in the sample. Bound haptoglobin is visualized with a specific anti-bovine haptoglobin antibody coupled to an enzyme that generates a color signal.
For Veterinary Use Only.

Specifications

Assay Sensitivity: See product documentation
Gene ID: Non-Gene
Gene Symbol: Non-Gene
Immunoassay Kit Format: Various Other
Product Line: Bio-X, Prionics™
Product Size: 42 samples
Target (Kit Named): Bovine Haptoglobine
Target Species (Named): Bovine
Target Species (Validated): Bovine

Documents

Manuals & protocols