Regulatory requirements vary by country; products may not be available in your geographic area.

Enterotoxaemia (Alpha, Beta, Epsilon, C. perfringens) ELISA Kit

Catalog number: 5031020

 Related applications: Animal Health

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top


The Enterotoxaemia (Alpha, Beta, Epsilon, C. perfringens) ELISA Kit is a sandwich ELISA kit for the detection of Clostridium perfringens Alpha, Beta, and Epsilon toxins and the bacterium in biological fluids or culture supernatants. Specific monoclonal and polyclonal antibodies produced against the toxins and a monoclonal antibody specific for a structural protein of the bacterium have been immobilized on alternate rows of microtiter plates. These antibodies allow the capture of the corresponding toxins or bacteria present in the sample.

Samples are incubated in the microtiter plate and bound toxins or proteins are detected using anti-pathogen monoclonal or polyclonal antibodies coupled to an enzyme that generates a color signal. The intensity of the resulting blue color is proportionate to the specific pathogen titer in the sample.
For Veterinary Use Only.


Species: All Sensitive Animals
Product Size: 24 samples
Protein Family: Other Proteins
Detection Method: Colorimetric


Manuals & protocols