Interested in a bulk or custom version of this product? Request pricing.

ECM1 Recombinant Mouse Protein, His Tag

Catalog number: 50331M08H25

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Extracellular Matrix Protein 1 (Ecm1), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

This recombinant protein was expressed from a DNA sequence encoding the mouse ECM1 isoform 1 (Q61508-1) (Met 1-Glu 559) fused to a polyhistidine tag at the C-terminus.

Activity: Measured by its ability to bind human MMP-9 in a functional ELISA.

Formulation: Lyophilized in 140 mM NaCl, 2.7 mM KCl, 10 mM Na2HPO4, 1.8 mM KH2PO4, pH7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.

This protein is manufactured by Sino Biological Inc.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Endotoxin Level: < 1 EU/µg
Expression: Mammalian
Form: Lyophilized
Gene Aliases: AI663821, p85
Gene Symbol: Ecm1
Molecular Weight: 62.5 kDa
Product Line: Sino Biological®
Product Size: 5 x 5 µg
Protein Family: Other Proteins
Protein Name: Ecm1 extracellular matrix protein 1
Protein Subtype: Glycoproteins
Protein Tag or Fusion: His Tag
Purity: ≥ 85% by SDS-PAGE
Research Category: Angiogenesis, Bone Research, Cell Cycle & Proliferation, Inflammation, Oncology, Other Research
Species: Mouse
Validated Application: Assay Standard

Contents & storage

Includes 5 vial of lyophilized protein. Store at -20 °C.

Documents

Manuals & protocols