SRD-3x00 Solvent Racks with Degassers for UltiMate™ 3000 Pumps
Actual product may vary
SRD-3x00 Solvent Racks with Degassers for UltiMate™ 3000 Pumps
SRD-3x00 Solvent Racks with Degassers for UltiMate™ 3000 Pumps
SRD-3x00 Solvent Racks with Degassers for UltiMate™ 3000 Pumps
Thermo Scientific™

SRD-3x00 Solvent Racks with Degassers for UltiMate™ 3000 Pumps

Enjoy stable baselines while securing multiple eluents in a high-capacity solvent rack. Supports high-pressure gradient (HPG) pumps without internal degassers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSolvent Channels
5035.92306 Degasser Channels
5035.92454 Degasser Channels
5035.92502 Degasser Channels
Catalog number 5035.9230
Price (MXN)
-
Solvent Channels:
6 Degasser Channels

Ensure stable baselines and consistent detector performance. The Thermo Scientific™ Dionex™ SRD-3x00 Solvent Racks Solvent Racks with Degassers for UltiMate™ 3000 Pumps (SRD-3200, -3400, and -3600) incorporate a low-volume (670μL), chemically-inert degasser with two, four, or six channels, respectively.

The SRD-3200 Solvent Rack with Degasser supports isocratic (ISO-3100SD) and high-pressure gradient (HPG-3200RS and HPG-3200SD) pumps which do not have built-in degassers. The SRD-3400 Solvent Rack with Degasser supports nano and capillary flow (NCS3500RS) and HPG (HPG-3400RS and HPG-3400SD) pumps without degassers. The SRD-3600 Solvent Rack with Degasser supports dual ternary rapid separation (DGP-3600RS), dual-gradient analytical (DGP-3600SD), and biocompatible dual-gradient micro (DGP-3600BM) pumps without degassers. An optional upgrade kit adds one more channel to degas the autosampler solvent to maintain reproducibility.

Features
  • Highest degassing efficiency ensures stable baselines.
  • Secure rack organizes up to nine 1-L eluent bottles on top of your Dionex UltiMate 3000 system tower.
  • Footprint conserves valuable bench space.
  • Resistance to the most commonly used high-performance liquid chromatography (HPLC) solvents and buffer solutions enables easy cleaning and maintenance.
  • Vacuum/leak sensors ensure continued safe operation.
  • Upgrade kit is available for added degassing channels.
Specifications
DescriptionSRD-3600 Solvent Rack
For Use With (Equipment)UltiMate 3000 Pumps
Voltage30 V
Weight (Metric)4.8 kg
Width (Metric)3 kg
Solvent Channels6 Degasser Channels
TypeSolvent Rack
Unit SizeEach