Search
Search

| Catalog Number | Diameter (Metric) | Quantity |
|---|---|---|
| 5036A | 0.36 μm | 15 mL |
| 5031A | 0.31 μm | 15 mL |
| 5031B | 0.31 μm | 100 mL |
| 5031C | 0.31 μm | 1000 mL |
| 5033A | 0.33 μm | 15 mL |
| 5033B | 0.33 μm | 100 mL |
| 5033C | 0.33 μm | 1000 mL |
| 5036B | 0.36 μm | 100 mL |
| 5036C | 0.36 μm | 1000 mL |
| 5043A | 0.43 μm | 15 mL |
| 5043B | 0.43 μm | 100 mL |
| 5043C | 0.43 μm | 1000 mL |
| 5049A | 0.49 μm | 15 mL |
| 5049B | 0.49 μm | 100 mL |
| 5049C | 0.49 μm | 1000 mL |
| 5050A | 0.5 μm | 15 mL |
| 5050B | 0.5 μm | 100 mL |
| 5050C | 0.5 μm | 1000 mL |
| 5051A | 0.51 μm | 15 mL |
| 5051B | 0.51 μm | 100 mL |
| 5051C | 0.51 μm | 1000 mL |
| 5052A | 0.52 μm | 15 mL |
| 5052B | 0.52 μm | 100 mL |
| 5060A | 0.6 μm | 15 mL |
| 5060B | 0.6 μm | 100 mL |
| 5060C | 0.6 μm | 1000 mL |
| 5067A | 0.67 μm | 15 mL |
| 5067B | 0.67 μm | 100 mL |
| 5067C | 0.67 μm | 1000 mL |
| 5074A | 0.75 μm | 15 mL |
| 5074B | 0.75 μm | 100 mL |
| 5074C | 0.75 μm | 1000 mL |
| 5081A | 0.82 μm | 15 mL |
| 5081B | 0.82 μm | 100 mL |
| 5081C | 0.82 μm | 1000 mL |
| 5088A | 0.87 μm | 15 mL |
| 5088B | 0.87 μm | 100 mL |
| 5088C | 0.87 μm | 1000 mL |
| 5093A | 0.93 μm | 15 mL |
| 5093B | 0.93 μm | 100 mL |
| 5045A | 0.45 μm | 15 mL |
| 5045C | 0.45 μm | 1000 mL |
| 5052C | 0.52 μm | 1000 mL |
| 5045B | 0.45 μm | 100 mL |
| 5093C | 0.93 μm | 1000 mL |
Features:
Ordering Information:
Includes:
Material Safety Data Sheet (MSDS) and general product handling insert sheet
Recommended for:
High concentration polymer particles < 3μm used in filter evaluation and testing, fluid mechanics research, dispersion studies and many other research and development projects. Not intended for use in instrument calibration or diagnostic reagents because they lack the exacting specifications needed for those applications.