Interested in a bulk or custom version of this product? Request pricing.

CD80 Recombinant Mouse Protein, His Tag

Catalog number: 50446M08H25

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

CD80 antigen (CD80), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of mouse CD80 (Q00609-1) (Met 1-Lys 245) fused to a polyhistidine tag at the C-terminus.

Activity: Measured by its ability to bind recombinant mouse CD28 in a functional ELISA.

Formulation: Lyophilized in 140 mM NaCl, 2.7 mM KCl, 10 mM Na2HPO4, 1.8 mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.

This protein is manufactured by Sino Biological Inc.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Protein Family: CD Proteins
Research Category: Cell Cycle & Proliferation, Immunology
Endotoxin Level: < 1 EU/µg
Expression: Mammalian
Form: Lyophilized
Gene Aliases: B71, Cd28l, Ly-53, Ly53, MIC17, TSA1
Gene Symbol: CD80
Molecular Weight: 25.1 kDa
Product Line: Sino Biological®
Product Size: 5 x 5 µg
Protein Name: CD80 Antigen
Protein Subtype: Other Proteins
Protein Tag or Fusion: His Tag
Purity: ≥ 90% by SDS-PAGE
Species: Mouse
Validated Application: Assay Standard

Contents & storage

Includes 5 vial of lyophilized protein. Store at -20 °C.

Documents

Manuals & protocols