Cryo Vial Closure Color Coders
Cryo Vial Closure Color Coders
Cryo Vial Closure Color Coders
Cryo Vial Closure Color Coders
Thermo Scientific™
Cryo Vial Closure Color Coders
Make it easier to identify specific vials quickly and simplify your inventory by using these color coders that fit into the top of cryogenic vial or tube closures.
Have Questions?
Alterar visualizaçãobuttonViewtableView
Número do catálogoColorProduct Line
5045-0002YellowNalgene
354755WhiteNunc
354879BlueNunc
354968RedNunc
355018GreenNunc
355077YellowNunc
355158OrangeNunc
375868BrownNunc
375884PinkNunc
375906GrayNunc
375922PurpleNunc
375930AssortedNunc
5045-0003BlueNalgene
5045-0000WhiteNalgene
5045-0004GreenNalgene
5045-0005RedNalgene
Número do catálogo 5045-0002
Preço (BRL)
-
Color:
Yellow
Product Line:
Nalgene
Make it possible to identify specific vials quickly and simplify your inventory using color coders that fit into the top of Nalgene™ Cryogenic vial closures or Nunc™ Cryogenic Tube closures.
  • Nalgene™ color coders are made from polystyrene and fit only in Nalgene™ cryogenic vial closures. Available in 5 colors
  • Nunc™ color coders are made from polycarbonate and fit only in Nunc™ cryogenic tube closures and Cryobank 2ml and 5mL tube caps. Available in 10 colors or as an assortment.
Especificações
DescriptionNalgene™ Color coder
Product TypeVial Closure
ColorYellow
MaterialPolycarbonate
Product LineNalgene
Quantity100/Pk., 500/Cs.
Unit SizeCase of 500