Cryo Vial Closure Color Coders
Cryo Vial Closure Color Coders
Cryo Vial Closure Color Coders
Cryo Vial Closure Color Coders
Thermo Scientific™
Cryo Vial Closure Color Coders
Make it easier to identify specific vials quickly and simplify your inventory by using these color coders that fit into the top of cryogenic vial or tube closures.
Have Questions?
變更視圖buttonViewtableView
產品號碼ColorProduct LineUnit Size
5045-0003PK
亦稱為 5045-0003
BlueNalgenePack of 100
354755WhiteNuncCase of 2000
354755PK
亦稱為 354755
WhiteNuncPack of 500
354879BlueNuncCase of 2000
354879PK
亦稱為 354879
BlueNuncPack of 500
354968RedNuncCase of 2000
354968PK
亦稱為 354968
RedNuncPack of 500
355018GreenNuncCase of 2000
355018PK
亦稱為 355018
GreenNuncPack of 500
355077YellowNuncCase of 2000
355077PK
亦稱為 355077
YellowNuncPack of 500
355158OrangeNuncCase of 2000
355158PK
亦稱為 355158
OrangeNuncPack of 500
375868BrownNuncCase of 2000
375868PK
亦稱為 375868
BrownNuncPack of 500
375884PinkNuncCase of 2000
375884PK
亦稱為 375884
PinkNuncPack of 500
375906GrayNuncCase of 2000
375906PK
亦稱為 375906
GrayNuncPack of 500
375922PurpleNuncCase of 2000
375922PK
亦稱為 375922
PurpleNuncPack of 500
375930AssortedNuncCase of 2000
375930PK
亦稱為 375930
AssortedNuncPack of 500
5045-0003BlueNalgeneCase of 500
5045-0000WhiteNalgeneCase of 500
5045-0000PK
亦稱為 5045-0000
WhiteNalgenePack of 100
5045-0002YellowNalgeneCase of 500
5045-0002PK
亦稱為 5045-0002
YellowNalgenePack of 100
5045-0004GreenNalgeneCase of 500
5045-0004PK
亦稱為 5045-0004
GreenNalgenePack of 100
5045-0005RedNalgeneCase of 500
5045-0005PK
亦稱為 5045-0005
RedNalgenePack of 100
產品號碼 5045-0003PK
亦稱為 5045-0003
價格 (HKD)
-
Color:
Blue
Product Line:
Nalgene
Unit Size:
Pack of 100
Make it possible to identify specific vials quickly and simplify your inventory using color coders that fit into the top of Nalgene™ Cryogenic vial closures or Nunc™ Cryogenic Tube closures.
  • Nalgene™ color coders are made from polystyrene and fit only in Nalgene™ cryogenic vial closures. Available in 5 colors
  • Nunc™ color coders are made from polycarbonate and fit only in Nunc™ cryogenic tube closures and Cryobank 2ml and 5mL tube caps. Available in 10 colors or as an assortment.
規格
DescriptionNalgene™ Color coder
Product TypeVial Closure
ColorBlue
MaterialPolycarbonate
Product LineNalgene
Quantity100/Pk., 500/Cs.
Unit SizePack of 100