Interested in a bulk or custom version of this product? Request pricing.

PDGFC Recombinant Mouse Protein, hIgG1-Fc Tag

Catalog number: 50458M01H25

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Platelet-derived growth factor C (PDGFC), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.

This recombinant protein was expressed from a DNA sequence encoding the receptor-binding form of mouse PDGFC (Q8CI19) (Val 235-Gly 345) fused to the Fc region of human IgG1 at the N-terminus.

Activity:
1. Measured by its binding ability in a functional ELISA. Immobilized mouse PDGFC (100 µl/well of a 10 µg/mlsolution) can bind human PDGFRA.
2. Measured in a cell proliferation assay using BALB/c 3T3 mouse embryonic fibroblast cells. The EC50 for this effect is typically 0.5-2 µg/ml.

Formulation: Lyophilized in 140 mM NaCl, 2.7 mM KCl, 10 mM Na2HPO4, 1.8 mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.



This protein is manufactured by Sino Biological Inc.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Research Category: Angiogenesis, Cardiovascular Research, Drug Development, Other Research, Signal Transduction
Endotoxin Level: < 1 EU/µg
Expression: Mammalian
Form: Lyophilized
Gene Aliases: 1110064L01Rik, AI647969, PDGF-C
Gene Symbol: PDGFC
Molecular Weight: 40.9 kDa
Product Line: Sino Biological®
Product Size: 5 x 5 µg
Protein Family: Growth Factors & Receptors
Protein Form: Recombinant, Truncated
Protein Name: platelet-derived growth factor, C polypeptide
Protein Subtype: PDGFs (Platelet-Derived Growth Factors)
Protein Tag or Fusion: IgG1 Fc Tag
Purity: ≥ 90% by SDS-PAGE
Species: Mouse
Validated Application: Assay Standard

Contents & storage

Includes 5 vial of lyophilized protein. Store at -20 °C.

Documents

Manuals & protocols

Recommended products