Interested in a bulk or custom version of this product? Request pricing.

CTLA4 Recombinant Mouse Protein, His Tag

Catalog number: 50503M08H25

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Cytotoxic T-Lymphocyte-Associated 4 (CTLA4), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of mouse CTLA4 (NP_033973.2) (Met 1-Phe 162) fused to a polyhistidine tag at the C-terminus.

Activity: Measured by its binding ability in a functional ELISA.
1. Immobilized recombinant mouse CTLA4-His (Cat:50503-M08H) at 10 µg/ml (100 µl/well) can mouse CD80-Fc (cat:50446-M02H) with a linear range of 0.78-62.5 ng/ml.
2. Immobilized recombinant mouse CTLA4-His (Cat:50503-M08H) at 10 µg/ml (100 µl/well) can human B7-1-Fc (Cat:10698-H03H) with a linear range of 0.78-125 ng/ml..

Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

This protein is manufactured by Sino Biological Inc.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Protein Family: CD Proteins
Research Category: Immunology, Signal Transduction
Endotoxin Level: < 1 EU/µg
Expression: Mammalian
Form: Lyophilized
Gene Aliases: CH29-120A16.2, Cd152, Ctla-4, Ly-56, Ctla4
Gene ID: 12477
Gene Symbol: CTLA4
Molecular Weight: 15.3 kDa
Product Line: Sino Biological®
Product Size: 5 x 5 µg
Protein Form: Recombinant, Truncated
Protein Name: Cytotoxic T-lymphocyte-associated protein 4
Protein Tag or Fusion: His Tag
Purification: PAGE
Purity: ≥ 90% by SDS-PAGE
Species: Mouse
Tag Position: C-Terminal
Validated Application: Assay Standard

Contents & storage

Includes 5 vials of protein in suspension buffer. Store at -20 °C..

Documents

Manuals & protocols

Recommended products