Interested in a bulk or custom version of this product? Request pricing.

GAD2 (GAD65) Recombinant Mouse Protein, N-His.GST Tag

Catalog number:  50653M20B25

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Glutamate Decarboxylase 2 (pancreatic islets and brain 65kDa) (GAD2), recombinant mouse protein is supplied in frozen format. In general, recombinant proteins can be used as protein standard and in cell biology research applications.

This recombinant protein was expressed from a DNA sequence encoding full length of mouse GAD2 (NP_032104.2) (Met 1-Leu 585) fused to the N-terminal polyhistidine-tagged GST tag at the N-terminus.

Activity: This recombinant protein has not been tested.

Formulation: 50 mM Tris, 100 mM NaCl, 0.5 mM PMSF, 0.5 mM GSH, 3 mM DTT, 10% glycerol, pH 8.0.

Reconstitution: No reconstitution needed. Store at -80°C. Thaw protein on ice before use and avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.

This protein is manufactured by Sino Biologica Inc.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Protein Family: Other Proteins
Research Category: Endocrinology
Endotoxin Level: < 1 EU/µg
Expression: Insect
Form: Frozen
Gene Aliases: Gad2, RP23-407K8.5, 6330404F12Rik, GAD(65), GAD65, Gad-2
Gene ID: 14417
Gene Symbol: GAD2
Molecular Weight: 93 kDa
Product Line: Sino Biological®
Product Size: 5 x 5 µg
Protein Form: Full Length, Recombinant
Protein Name: glutamate decarboxylase 2 (pancreatic islets and brain, 65kDa)
Protein Tag or Fusion: GST Tag, His Tag
Purification: PAGE
Purity: ≥ 90% by SDS-PAGE
Species: Mouse
Validated Application: Assay Standard

Contents & storage

Includes 5 vials of protein in suspension buffer. Store at -80 °C.

Documents

Manuals & protocols