Interested in a bulk or custom version of this product? Request pricing.

UCHL1 (UCH-L1) Recombinant Mouse Protein, N-His Tag

Catalog number: 50690M07E25

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Ubiquitin Carboxy-terminal Hydrolase L1 (UCHL1), recombinant mpuse protein is supplied as a frozen protein. It is suitable for use in protein studies such as enzymetic analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

This recombinant protein was expressed from a DNA sequence encoding the mouse UCHL1 (Q9R0P9) (Gln 2-Ala 223) fused to a polyhistide tag at the N-terminus.

Activity: Measured by the hydrolysis of UbiquitinAMC. The specific activity is >14000 pmoles/min/µg.

Formulation: 50 mM Tris, 150 mM NaCl, 20% Glycerol, PH7.7, 5% Mannitol, 5% Trehalose, 0.02% Tween80.

Reconstitution: No reconstitution needed. Store at -80°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.



This protein is manufactured by SINO Biological.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Protein Family: Other Proteins
Research Category: Neurobiology, Oncology
Expression: E. coli
Form: Frozen, Lyophilized
Gene Aliases: AW822034, C88048, PGP9.5, R75593, gad
Gene ID: 22223
Gene Symbol: UCHL1
Molecular Weight: 26.2 kDa
Product Line: Sino Biological®
Product Size: 5 x 5 µg
Protein Form: Full Length, Recombinant
Protein Name: Ubiquitin Carboxy-terminal Hydrolase L1, Ubiquitin carboxyl-terminal esterase L1
Protein Subtype: Proteases
Protein Tag or Fusion: His Tag
Purification: PAGE
Purity: ≥ 90% by SDS-PAGE
Species: Mouse
Tag Position: N-Terminal
Validated Application: Assay Standard

Contents & storage

Includes 5 vials of protein in suspension buffer. Store at -80 °C.

Documents

Manuals & protocols