Description | FLD-3400 RS Rapid Separation Fluorescence Detector with Dual-PMT, four channels, without flow cell |
Data Collection Rate | 200 Hz |
Detection Mode | 4 Channels |
Detector Type | High-Sensitivity Fluorescence |
Type | Fluorescence Detector |
Display | LCD Monitor |
Filter | Variable |
Flow Cell Max. Pressure | 290 psi |
Flow Cell Thermostatting | 15°C Above Ambient -50°C |
Flow Cell Volume | 8 μL |
Flow Rate | Analytical to Semi-Prep |
GLP Features | Automatic Instrument Qualification (AutoQ), System Wellness and Qualification Monitoring with Chromeleon Software, Lamp and Cell ID Chips. |
Lamp | Xenon Flash Lamp |
Lamp Pulse Frequency | 300 Hz |
No. of Channels | 4 |
PC Connection | USB |
Sensitivity | >550 ASTM |
Spectral Bandwidth | 20 nm |
Temperature Control | Yes |
Wavelength Accuracy | ±2 nm |
Wavelength Range | 200 to 880 nm |
Wavelength Repeatability | ±0.2 nm |
Wavelength Switching Time | <250 ms |
Weight (Metric) | 18 kg |
Unit Size | Each |
Description | FLD-3400 RS Rapid Separation Fluorescence Detector, four channels, without flow cell |
Data Collection Rate | 200 Hz |
Detection Mode | 4 Channels |
Detector Type | High-Sensitivity Fluorescence |
Type | Fluorescence Detector |
Display | LCD Monitor |
Filter | Variable |
Flow Cell Max. Pressure | 290 psi |
Flow Cell Thermostatting | 15°C Above Ambient -50°C |
Flow Cell Volume | 8 μL |
Flow Rate | Analytical to Semi-Prep |
GLP Features | Automatic Instrument Qualification (AutoQ), System Wellness and Qualification Monitoring with Chromeleon Software, Lamp and Cell ID Chips. |
Lamp | Xenon Flash Lamp |
Lamp Pulse Frequency | 300 Hz |
No. of Channels | 4 |
PC Connection | USB |
Sensitivity | >550 ASTM |
Spectral Bandwidth | 20 nm |
Temperature Control | Yes |
Wavelength Accuracy | ±2 nm |
Wavelength Range | 200 to 630 nm |
Weight (Metric) | 18 kg |
Unit Size | Each |
Description | FLD-3100 Fluorescence Detector with Dual-PMT, one channel, without flow cell |
Data Collection Rate | 100 Hz |
Detection Mode | Single Channel |
Detector Type | High-Sensitivity Fluorescence |
Type | Fluorescence Detector |
Display | LCD Monitor |
Filter | Fixed |
Flow Cell Max. Pressure | 290 psi |
Flow Cell Thermostatting | 15°C Above Ambient -50°C |
Flow Cell Volume | 8 μL |
GLP Features | Automatic Instrument Qualification (AutoQ), System Wellness and Qualification Monitoring with Chromeleon Software, Lamp and Cell ID Chips. |
Lamp | Xenon Flash Lamp |
Lamp Pulse Frequency | 300 Hz |
No. of Channels | 1 |
PC Connection | USB |
Sensitivity | >550 ASTM |
Spectral Bandwidth | 20 nm |
Temperature Control | Yes |
Wavelength Accuracy | ±2 nm |
Wavelength Range | 200 to 880 nm |
Weight (Metric) | 18 kg |
Unit Size | Each |
Description | FLD-3100 Fluorescence Detector, one channel, without flow cell |
Data Collection Rate | 100 Hz |
Detection Mode | Single Channel |
Detector Type | High-Sensitivity Fluorescence |
Type | Fluorescence Detector |
Display | LCD Monitor |
Filter | Fixed |
Flow Cell Max. Pressure | 290 psi |
Flow Cell Thermostatting | 15°C Above Ambient -50°C |
Flow Cell Volume | 8 μL |
GLP Features | Automatic Instrument Qualification (AutoQ), System Wellness and Qualification Monitoring with Chromeleon Software, Lamp and Cell ID Chips. |
Lamp | Xenon Flash Lamp |
Lamp Pulse Frequency | 300 Hz |
No. of Channels | 1 |
PC Connection | USB |
Sensitivity | >550 ASTM |
Spectral Bandwidth | 20 nm |
Temperature Control | Yes |
Wavelength Accuracy | ±2 nm |
Wavelength Range | 200 to 630 nm |
Weight (Metric) | 18 kg |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | Detection Mode | Price (USD) | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
5078.0025 | Each | FLD-3400 RS Rapid Separation Fluorescence Detector with Dual-PMT, four channels, without flow cell | 4 Channels | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
5078.0020 | Each | FLD-3400 RS Rapid Separation Fluorescence Detector, four channels, without flow cell | 4 Channels | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
5078.0015 | Each | FLD-3100 Fluorescence Detector with Dual-PMT, one channel, without flow cell | Single Channel | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
5078.0010 | Each | FLD-3100 Fluorescence Detector, one channel, without flow cell | Single Channel | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
The Thermo Scientific™ UltiMate 3000 Rapid Separation Fluorescence Detector FLD-3400 is recommended for method development labs that need flexibility in detecting novel compounds that don’t absorb in the UV-vis region of the spectrum. It is ideal for the analysis of carbohydrates, glycans, and polyaromatic hydrocarbons (PAHs) in combination with any Thermo Scientific™ Dionex™ UltiMate 3000 system. The ability to cleave and derivitize glycans from intact proteins or monoclonal antibodies eliminates interferences from coeluting matrix peptides.
Sensitive Fluorescence Performance
Easy-to-Use Fluorescence Detector