Thermo Scientific™ QSP™ Snap Cap Microcentrifuge Tube
Thermo Scientific™
Thermo Scientific™ QSP™ Snap Cap Microcentrifuge Tube
Thermo Scientific™ QSP™ Snap Cap Microcentrifuge Tube withstands the rigors of sample preparation, transportation and short term storage, simplfying the laboratory process with a single tube.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorSterilityMax. RCF
508-GRD-G-QGreenNon-sterile30,000 x g
502-GRD-V-QVioletNon-sterile30,000 x g
502-GRD-R-QRedNon-sterile30,000 x g
508-GRD-A-QAmberNon-sterile30,000 x g
502-GRD-Y-QYellowNon-sterile30,000 x g
502-GRD-O-QOrangeNon-sterile30,000 x g
509-GRD-G-QGreenNon-sterile30,000 x g
509-GRD-Y-QYellowNon-sterile30,000 x g
502-GRD-B-QBlueNon-sterile30,000 x g
502-GRDS-QNaturalSterile30,000 x g
509-GRD-R-QRedNon-sterile30,000 x g
508-GRD-R-QRedNon-sterile30,000 x g
508-GRD-B-QBlueNon-sterile30,000 x g
509-GRD-A-QAmberNon-sterile30,000 x g
502-GRD-A-QAmberNon-sterile30,000 x g
509-GRDS-QNaturalSterile30,000 x g
509-GRD-O-QOrangeNon-sterile30,000 x g
508-GRD-V-QVioletNon-sterile30,000 x g
502-GRD-MIXED-QAssortedNon-sterile30,000 x g
508-GRDS-QNaturalSterile30,000 x g
508-GRD-Y-QYellowNon-sterile30,000 x g
508-GRD-O-QOrangeNon-sterile30,000 x g
508-GRD-MIXED-QAssortedNon-sterile30,000 x g
509-GRD-MIXED-QAssortedNon-sterile30,000 x g
509-GRD-B-QBlueNon-sterile30,000 x g
502-GRD-G-QGreenNon-sterile30,000 x g
509-GRD-V-QVioletNon-sterile30,000 x g
Catalog number 508-GRD-G-Q
Price (TWD)
-
Color:
Green
Sterility:
Non-sterile
Max. RCF:
30,000 x g
Thermo Scientific™ QSP™ Snap Cap Microcentrifuge Tube 0.6ml Graduated at 0.2, 0.4, 0.6ml, withstand up to 30,000 x g, frosted tube and cap and Amber color. Non-sterile clean claims; Rnase/Dnase, DNA, pyrogen and endotoxin free.
  • Volume range from 0.5ml to 2.0ml, available in sterile and non sterile options
  • Graduation marks
  • Colored options
  • Snap cap tube easy to open and close withstanding force up to 30,000 x G
Specifications
Clean ClaimsNon-Sterile free of: Rnase/Dnase, DNA, pyrogen and endotoxin
AutoclavableAutoclavable
ColorGreen
GraduatedYes
MaterialPolypropylene (Virgin)
Max. RCF30,000 x g
SterilityNon-sterile
Unit SizeCase of 5000