Interested in a bulk or custom version of this product? Request pricing.

PTPN2 Recombinant Mouse Protein, N-His Tag

Catalog number: 50800M07B5

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Protein Tyrosine Phosphatase Non-Receptor Type 2 (PTPN2), recombinant mouse protein is supplied in frozen format. It is suitable for use in protein studies such as enzymetic analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

This recombinant protein was expressed from a DNA sequence encoding the amino acid (Met 1-Asn 314) of mouse PTPN2 (P17706-1) fused to the N-terminal polyhistidine-tagged GST tag at the N-terminus.

Activity: Measured by its ability to dephosphorylate a phosphotyrosine residue in an EGF receptor (988-998 phosphopeptide substrate, R&D Systems, Catalog # ES006).The specific activity is>15 nmoles/min/µg.

Formulation: 20 mM Tris,500 mM NaCl, 0.2 µg/µl BSA, pH 8.0, 20% glycerol, 0.1 mM TCEP.

Reconstitution: No reconstitution needed. Store at -80°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.

This protein is manufactured by Sino Biological Inc.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Endotoxin Level: < 1 EU/µg
Expression: Insect
Form: Frozen
Gene Aliases: AI325124, Ptpt, TC-PTP
Gene Symbol: PTPN2
Molecular Weight: 38.7 kDa
Product Line: Sino Biological®
Product Size: 5 µg
Protein Name: protein tyrosine phosphatase, non-receptor type 2
Protein Subtype: Tyrosine Phosphatases
Protein Tag or Fusion: His Tag
Purity: ≥ 85% by SDS-PAGE
Research Category: Drug Development, Other Research
Species: Mouse
Validated Application: Assay Standard

Contents & storage

Store at -80 °C.

Documents

Manuals & protocols