Rat IgG2a kappa Isotype Control (eBR2a), Alexa Fluor™ 647, eBioscience™ - FAQs
Rat IgG2a kappa Isotype Control (eBR2a), Alexa Fluor™ 647, eBioscience™ - FAQs
View additional product information for Rat IgG2a kappa Isotype Control (eBR2a), Alexa Fluor™ 647, eBioscience™ - FAQs (51-4321-81)