Product Image
Thermo Scientific™

Cytomat™ 2C70-LIN Automated Incubator

Temperature Range: Ambient +10°C to 70°C; Relative Humidity: <95%
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVoltage
51022042100 V
51019061230 V
51020860120 V
Catalog number 51022042
Price (USD)
-
Voltage:
100 V

The Thermo Scientific™ Cytomat™ 2 series automated incubator is the most compact system with a capacity for 42 standard microplates. This series offers several models with various temperature ranges and gate positions (bottom, top). The standard models are bench top and space-saving under bench solutions are also available. Its design and unsurpassed temperature uniformity and stability ensure that your processes can be fully reproduced, providing you with the same high quality results every time.

The Cytomat 2 C70-LIN automated incubator provides an ideal environment for storage and incubation of assays in a temperature range of ambient +10° to 70°C, with high humidity up to 95% r.H. and stable CO2 conditions.

It offers extremely uniform environmental conditions for all microplates, elimination of recovery times, and reduced assay variance.

Matches your Capacity with 2 Stacker
  • 56pcs. 1536-well plates
  • 42pcs. 96-well or 384-well plates
  • 30pcs. transwell or insert plates
  • 20pcs. deep-well

Further stacker types on request
Mix and match different pitch stackers


Stable Humidity Condition
  • Specially designed internal water reservoir creates a stable, high level of humidity environment
  • Ventilated stackers minimize evaporation

Access Time
  • Below 20sec.

Contamination Prevention
  • Easy to clean - smooth stainless steel surface
  • Solid copper inner chamber (option)
  • H2O2-flange (Option)

Flexible Stacker Design
  • Compatible with existing Cytomat stacker
  • Standard or customized stacker pitch sizes
  • Mix and match different pitch stackers
  • Autoclaveable, stainless steel
  • Allows for accurate temperature distribution with ventilation slots

Convenient
  • Internal and direct barcode reader can scan barcodes at any plate position inside the incubator the incubator, minimizing plate movement and provides superior sample and plate tracking

Easy to integrate
  • Intuitive software commands
  • SILA (Standard in Lab Automation) Compliant
  • Direct connection to a clean bench
  • Multiple transfer stations

Certification
  • CE
  • RoHS
  • Manufactured in ISO 9001 and ISO 13485 accredited facility
Specifications
Access TimeBelow 20 sec.
Certifications/ComplianceCE Marked
Depth (English)17 in.
Depth (Metric)431 mm
Electrical Requirements100 V, 50 Hz
FeaturesIncubation and Storage Unit with Temperature Range Above, Humidity up to 95% r.H. and CO2 Range of 0-20 Vol. %.
Height (English)43.3 in.
Height (Metric)1100 mm
Holds56 Pc. 1536-Well Plates, 42 Pc. 96-Well or 384-Well Plates, 30 Pc. Transwell or Insert Plates, 20 Pc. Deep-Well
ModelCytomat 2 C70-LIN
TypeAutomated Incubator
Weight (Metric)80 kg
Width (English)19.3 in.
Width (Metric)491 mm
Chamber MaterialCopper
Relative Humidity<95%
Temperature Range (Metric)Ambient +10° to 70°C
Voltage100 V
Unit SizeEach