Cytomat™ 10 C Series Automated Incubators
Actual product may vary
Thermo Scientific™

Cytomat™ 10 C Series Automated Incubators

Temperature Range: Ambient +10°C to 50°C; Relative Humidity: <98% at 37°C
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVoltage
51025982120 V
51026519100 V
51025983230 V
Catalog number 51025982
Price (EUR)
-
Voltage:
120 V

Designed to fit into the busiest of cell biology or screening laboratories, the Thermo Scientific™ Cytomat™ 10 Automated Incubator offers superior environmental control and can be seamlessly integrated with any existing automated system.

The Cytomat 10 is a fully automated incubator and storage module for high-capacity cell growth and assay incubation in a temperature range of ambient +10°C to 50°C combined with controlled humidity up to 98% r.H. and stable CO2 conditions.

The Cytomat 10 provides a fast plate access time of less than 10 seconds. Additionally, multiple transfer positions and an under-bench version allow flexible incubator placement within the laboratory.

As the first generation of automated incubator with a fully automated thermal decontamination system, the Cytomat 10 is designed to significantly decrease the risk of contamination and reduce assay variability.

5 Stacker to Match Your Capacity

  • 140pcs. 1536-well plates
  • 105pcs. 96-well or 384-well plates
  • 75pcs. transwell or insert plates
  • 50pcs. deep-well plates

Mix and match different pitch stackers. Additional stacker types are available upon request.

Effortless Humidity Control

  • HydraSmart technology provides ultrasonic vapor technology with no turbulence and heat introduction into system
  • Sterilized water supply by external accessible micro-particle filter
  • Closed loop water delivery system, with simple external tank
  • Water level monitoring with refill warning

Fast

  • Below 10 second access time
  • Application speed settings: slow, medium, fast
  • Individual microplate speed settings

Contamination Prevention

  • ContraCon for an automated decontamination routine with true walk-away time
  • Easy to clean: smooth stainless steel surface
  • Solid copper inner chamber (optional)
  • H2O2 flange (optional)

Flexible Stacker Design

  • Compatible with existing Cytomat stacker
  • Standard or customized stacker pitch sizes
  • Mix and match different pitch stackers
  • Autoclaveable stainless steel
  • Ventilation slots allow accurate temperature distribution

Convenient, Easy to Use

  • 5-inch touch screen with LED backlight as standard
  • Various gate / transfer positions

Easy to integrate

  • Intuitive software commands
  • Flat gate, no gate cover
  • Designed with multiple transfer stations

More Options

  • Additional capacity: one additional stacker position on turntable
  • O2 control
  • Solid copper inner chamber
  • H2O2 flange
  • Internal and direct barcode reader

Certification

  • CE
  • RoHS III
  • Manufactured in ISO 9001 and ISO 13485 accredited facility
Specifications
Access TimeBelow 10 sec.
Certifications/ComplianceCE Marked
Depth (English)32.3 in.
Depth (Metric)822 mm
Electrical Requirements120 V, 60 Hz
FeaturesIncubation and Storage Unit with Temperature Range Below, Humidity up to 98% r.H. (at 37°C and CO2 Range of 0-20 Vol. %)
Height (English)36.0 in.
Height (Metric)915 mm
Holds280 Pc. 1536-Well Plates, 210 Pc. 96-Well or 384-Well Plates, 150 Pc. Transwell or Insert Plates, 100 Pc. Deep-Well
ModelCytomat 10 C
TypeAutomated Incubator
Width (English)31.5 in.
Width (Metric)800 mm
Chamber MaterialCopper
Relative Humidity<98%
Temperature Range (Metric)Ambient +10° to 50°C
Voltage120 V
Unit SizeEach