Thermo Scientific™
Heratherm™ Advanced Protocol Security Ovens
Catalog number: 51028545
Thermo Scientific™
Heratherm™ Advanced Protocol Security Ovens
Catalog number: 51028545
Protect samples effectively with Thermo Scientific™ Heratherm™ Advanced Protocol Security Ovens, which feature an extra layer of security for applications where process reliability and security are paramount. A standard over-temperature alarm and an additional under-temperature alarm ensure sample stability. A door lock prevents disruption or tampering of samples, while a door alarm notifies the operator when the door is left open accidentally. Choose a stainless steel exterior for demanding pharmaceutical and clinical applications.
Catalog Number
51028545
Unit Size
Each
Capacity (Metric)
99 L
Type
Gravity Convection
Electrical Requirements
230 V 50/60 Hz
Material
Stainless Steel
Price (USD)
Availability
***
Full specifications
Quantity
-
Amperage13.3 A
Controller TypeProgrammable Microprocessor control with vacuum fluorescent display
Depth (English) Interior15.3 in.
Depth (Metric) Interior38.9 cm
Dimensions (L x W x H)22.2 x 25.2 x 32.3 in. (56.5 x 64 x 82 cm)
Frequency50/60 Hz
Height (English)32.3 in.
Height (English) Interior22.8 in.
Height (Metric)82 cm
Height (Metric) Interior58 cm
Length (English)22.2 in.
Length (Metric)56.5 cm
Temperature Stability±0.3°C
TimerWeekly / Real time / Hour
Wattage3100 w
Width (English) Interior17.2 in.
Width (Metric) Interior43.8 cm
Temperature (Metric)50°C to 330°C
Voltage230 V
Capacity (Metric)99 L
Dimensions (D x W x H) Interior15.3 x 17.2 x 22.8 in. (38.9 x 43.8 x 58 cm)
TypeGravity Convection
Electrical Requirements230 V 50/60 Hz
MaterialStainless Steel
Plug TypeCountry-specific
Width (English)25.2 in.
No. of Shelves2 supplied/16 max.
Width (Metric)64 cm
Unit SizeEach
Catalog Number
Unit Size
Each
Capacity (Metric)
61 L
Type
Gravity Convection
Electrical Requirements
120 V 60 Hz
Material
Coated Steel
Price (USD)
2,891.00
Your Price:
Availability
***
Full specifications
Quantity
Amperage14.4 A
Controller TypeProgrammable Microprocessor control with vacuum fluorescent display
Depth (English) Interior15.3 in.
Depth (Metric) Interior38.9 cm
Dimensions (L x W x H)22.2 x 20.9 x 28.3 in. (56.5 x 53 x 72 cm)
Frequency60 Hz
Height (English)28.3 in.
Height (English) Interior18.9 in.
Height (Metric)72 cm
Height (Metric) Interior48 cm
Length (English)22.2 in.
Length (Metric)56.5 cm
Temperature Stability±0.3°C
TimerWeekly / Real time / Hour
Wattage1730 w
Width (English) Interior12.9 in.
Width (Metric) Interior32.8 cm
Temperature (Metric)50°C to 330°C
Voltage120 V
Capacity (Metric)61 L
Dimensions (D x W x H) Interior15.3 x 12.9 x 18.9 in. (38.9 x 32.8 x 48 cm)
TypeGravity Convection
Electrical Requirements120 V 60 Hz
MaterialCoated Steel
Plug TypeNema 5-15
Width (English)20.9 in.
No. of Shelves2 supplied/39 max.
Width (Metric)53 cm
Unit SizeEach
Catalog Number
Unit Size
Each
Capacity (Metric)
61 L
Type
Gravity Convection
Electrical Requirements
230 V 50/60 Hz
Material
Coated Steel
Price (USD)
Availability
***
Full specifications
Quantity
-
Amperage7.9 A
Controller TypeProgrammable Microprocessor control with vacuum fluorescent display
Depth (English) Interior15.3 in.
Depth (Metric) Interior38.9 cm
Dimensions (L x W x H)22.2 x 20.9 x 28.3 in. (56.5 x 53 x 72 cm)
Frequency50/60 Hz
Height (English)28.3 in.
Height (English) Interior18.9 in.
Height (Metric)72 cm
Height (Metric) Interior48 cm
Length (English)22.2 in.
Length (Metric)56.5 cm
Temperature Stability±0.3°C
TimerWeekly / Real time / Hour
Wattage1810 w
Width (English) Interior12.9 in.
Width (Metric) Interior32.8 cm
Temperature (Metric)50°C to 330°C
Voltage230 V
Capacity (Metric)61 L
Dimensions (D x W x H) Interior15.3 x 12.9 x 18.9 in. (38.9 x 32.8 x 48 cm)
TypeGravity Convection
Electrical Requirements230 V 50/60 Hz
MaterialCoated Steel
Plug TypeCountry-specific
Width (English)20.9 in.
No. of Shelves2 supplied/13 max.
Width (Metric)53 cm
Unit SizeEach
Catalog Number
Unit Size
Each
Capacity (Metric)
99 L
Type
Gravity Convection
Electrical Requirements
208/240 V 60 Hz
Material
Coated Steel
Price (USD)
3,131.00
Your Price:
Availability
***
Full specifications
Quantity
Amperage14 A
Controller TypeProgrammable Microprocessor control with vacuum fluorescent display
Depth (English) Interior15.3 in.
Depth (Metric) Interior38.9 cm
Dimensions (L x W x H)22.2 x 25.2 x 32.3 in. (56.5 x 64 x 82 cm)
Frequency60 Hz
Height (English)32.3 in.
Height (English) Interior22.8 in.
Height (Metric)82 cm
Height (Metric) Interior58 cm
Length (English)22.2 in.
Length (Metric)56.5 cm
Temperature Stability±0.3°C
TimerWeekly / Real time / Hour
Wattage3360 w
Width (English) Interior17.2 in.
Width (Metric) Interior43.8 cm
Temperature (Metric)50°C to 330°C
Voltage208/240 V
Capacity (Metric)99 L
Dimensions (D x W x H) Interior15.3 x 17.2 x 22.8 in. (38.9 x 43.8 x 58 cm)
TypeGravity Convection
Electrical Requirements208/240 V 60 Hz
MaterialCoated Steel
Plug TypeNema 6-20
Width (English)25.2 in.
No. of Shelves2 supplied/16 max.
Width (Metric)64 cm
Unit SizeEach
Catalog Number
Unit Size
Each
Capacity (Metric)
99 L
Type
Gravity Convection
Electrical Requirements
230 V 50/60 Hz
Material
Coated Steel
Price (USD)
Availability
***
Full specifications
Quantity
-
Amperage13.5 A
Controller TypeProgrammable Microprocessor control with vacuum fluorescent display
Depth (English) Interior15.3 in.
Depth (Metric) Interior38.9 cm
Dimensions (L x W x H)22.2 x 25.2 x 32.3 in. (56.5 x 64 x 82 cm)
Frequency50/60 Hz
Height (English)32.3 in.
Height (English) Interior22.8 in.
Height (Metric)82 cm
Height (Metric) Interior58 cm
Length (English)22.2 in.
Length (Metric)56.5 cm
Temperature Stability±0.3°C
TimerWeekly / Real time / Hour
Wattage3100 w
Width (English) Interior17.2 in.
Width (Metric) Interior43.8 cm
Temperature (Metric)50°C to 330°C
Voltage230 V
Capacity (Metric)99 L
Dimensions (D x W x H) Interior15.3 x 17.2 x 22.8 in. (38.9 x 43.8 x 58 cm)
TypeGravity Convection
Electrical Requirements230 V 50/60 Hz
MaterialCoated Steel
Plug TypeCountry-specific
Width (English)25.2 in.
No. of Shelves2 supplied/16 max.
Width (Metric)64 cm
Unit SizeEach
Catalog Number
Unit Size
Each
Capacity (Metric)
168 L
Type
Gravity Convection
Electrical Requirements
208/240 V 60 Hz
Material
Coated Steel
Price (USD)
Availability
***
Full specifications
Quantity
-
Amperage14 A
Controller TypeProgrammable Microprocessor control with vacuum fluorescent display
Depth (English) Interior22.2 in.
Depth (Metric) Interior56.4 cm
Dimensions (L x W x H)29.1 x 25.2 x 36.2 in. (73.8 x 64 x 92 cm)
Frequency60 Hz
Height (English)36.2 in.
Height (English) Interior26.8 in.
Height (Metric)92 cm
Height (Metric) Interior68 cm
Length (English)29.1 in.
Length (Metric)73.8 cm
Temperature Stability±0.3°C
TimerWeekly / Real time / Hour
Wattage3360 w
Width (English) Interior17.2 in.
Width (Metric) Interior43.8 cm
Temperature (Metric)50°C to 330°C
Voltage208/240 V
Capacity (Metric)168 L
Dimensions (D x W x H) Interior22.2 x 17.2 x 26.8 in. (56.4 x 43.8 x 68 cm)
TypeGravity Convection
Electrical Requirements208/240 V 60 Hz
MaterialCoated Steel
Plug TypeNema 6-20
Width (English)25.2 in.
No. of Shelves2 supplied/19 max.
Width (Metric)64 cm
Unit SizeEach
Catalog Number
Unit Size
Each
Capacity (Metric)
168 L
Type
Gravity Convection
Electrical Requirements
230 V 50/60 Hz
Material
Coated Steel
Price (USD)
Availability
***
Full specifications
Quantity
-
Amperage13.5 A
Controller TypeProgrammable Microprocessor control with vacuum fluorescent display
Depth (English) Interior22.2 in.
Depth (Metric) Interior56.4 cm
Dimensions (L x W x H)29.1 x 25.2 x 36.2 in. (73.8 x 64 x 92 cm)
Frequency50/60 Hz
Height (English)36.2 in.
Height (English) Interior26.8 in.
Height (Metric)92 cm
Height (Metric) Interior68 cm
Length (English)29.1 in.
Length (Metric)73.8 cm
Temperature Stability±0.3°C
TimerWeekly / Real time / Hour
Wattage3100 w
Width (English) Interior17.2 in.
Width (Metric) Interior43.8 cm
Temperature (Metric)50°C to 330°C
Voltage230 V
Capacity (Metric)168 L
Dimensions (D x W x H) Interior22.2 x 17.2 x 26.8 in. (56.4 x 43.8 x 68 cm)
TypeGravity Convection
Electrical Requirements230 V 50/60 Hz
MaterialCoated Steel
Plug TypeCountry-specific
Width (English)25.2 in.
No. of Shelves2 supplied/19 max.
Width (Metric)64 cm
Unit SizeEach
Catalog Number
Unit Size
Each
Capacity (Metric)
61 L
Type
Gravity Convection
Electrical Requirements
120 V 60 Hz
Material
Stainless Steel
Price (USD)
2,874.00
Your Price:
Availability
***
Full specifications
Quantity
Amperage14.4 A
Controller TypeProgrammable Microprocessor control with vacuum fluorescent display
Depth (English) Interior15.3 in.
Depth (Metric) Interior38.9 cm
Dimensions (L x W x H)22.2 x 20.9 x 28.3 in. (56.5 x 53 x 72 cm)
Frequency60 Hz
Height (English)28.3 in.
Height (English) Interior18.9 in.
Height (Metric)72 cm
Height (Metric) Interior48 cm
Length (English)22.2 in.
Length (Metric)56.5 cm
Temperature Stability±0.3°C
TimerWeekly / Real time / Hour
Wattage1730 w
Width (English) Interior12.9 in.
Width (Metric) Interior32.8 cm
Temperature (Metric)50°C to 330°C
Voltage120 V
Capacity (Metric)61 L
Dimensions (D x W x H) Interior15.3 x 12.9 x 18.9 in. (38.9 x 32.8 x 48 cm)
TypeGravity Convection
Electrical Requirements120 V 60 Hz
MaterialStainless Steel
Plug TypeNema 5-15
Width (English)20.9 in.
No. of Shelves2 supplied/13 max.
Width (Metric)53 cm
Unit SizeEach
Catalog Number
Unit Size
Each
Capacity (Metric)
61 L
Type
Gravity Convection
Electrical Requirements
230 V 50/60 Hz
Material
Stainless Steel
Price (USD)
Availability
***
Full specifications
Quantity
-
Amperage6.1 A
Controller TypeProgrammable Microprocessor control with vacuum fluorescent display
Depth (English) Interior15.3 in.
Depth (Metric) Interior38.9 cm
Dimensions (L x W x H)22.2 x 20.9 x 28.3 in. (56.5 x 53 x 72 cm)
Frequency50/60 Hz
Height (English)28.3 in.
Height (English) Interior18.9 in.
Height (Metric)72 cm
Height (Metric) Interior48 cm
Length (English)22.2 in.
Length (Metric)56.5 cm
Temperature Stability±0.3°C
TimerWeekly / Real time / Hour
Wattage1810 w
Width (English) Interior12.9 in.
Width (Metric) Interior32.8 cm
Temperature (Metric)50°C to 330°C
Voltage230 V
Capacity (Metric)61 L
Dimensions (D x W x H) Interior15.3 x 12.9 x 18.9 in. (38.9 x 32.8 x 48 cm)
TypeGravity Convection
Electrical Requirements230 V 50/60 Hz
MaterialStainless Steel
Plug TypeCountry-specific
Width (English)20.9 in.
No. of Shelves2 supplied/13 max.
Width (Metric)53 cm
Unit SizeEach
Catalog Number
Unit Size
Each
Capacity (Metric)
99 L
Type
Gravity Convection
Electrical Requirements
208/240 V 60 Hz
Material
Stainless Steel
Price (USD)
Availability
***
Full specifications
Quantity
-
Amperage14 A
Controller TypeProgrammable Microprocessor control with vacuum fluorescent display
Depth (English) Interior15.3 in.
Depth (Metric) Interior38.9 cm
Dimensions (L x W x H)22.2 x 25.2 x 32.3 in. (56.5 x 64 x 82 cm)
Frequency60 Hz
Height (English)32.3 in.
Height (English) Interior22.8 in.
Height (Metric)82 cm
Height (Metric) Interior58 cm
Length (English)22.2 in.
Length (Metric)56.5 cm
Temperature Stability±0.3°C
TimerWeekly / Real time / Hour
Wattage3360 w
Width (English) Interior17.2 in.
Width (Metric) Interior43.8 cm
Temperature (Metric)50°C to 330°C
Voltage208/240 V
Capacity (Metric)99 L
Dimensions (D x W x H) Interior15.3 x 17.2 x 22.8 in. (38.9 x 43.8 x 58 cm)
TypeGravity Convection
Electrical Requirements208/240 V 60 Hz
MaterialStainless Steel
Plug TypeNema 6-20
Width (English)25.2 in.
No. of Shelves2 supplied/16 max.
Width (Metric)64 cm
Unit SizeEach
Showing 10 of 23Show more
Catalog NumberSpecificationsUnit SizeCapacity (Metric)TypeElectrical RequirementsMaterialPrice (USD)AvailabilityQuantity
51028545Full specifications
Each99 LGravity Convection230 V 50/60 HzStainless SteelRequest A Quote***-
Amperage13.3 A
Controller TypeProgrammable Microprocessor control with vacuum fluorescent display
Depth (English) Interior15.3 in.
Depth (Metric) Interior38.9 cm
Dimensions (L x W x H)22.2 x 25.2 x 32.3 in. (56.5 x 64 x 82 cm)
Frequency50/60 Hz
Height (English)32.3 in.
Height (English) Interior22.8 in.
Height (Metric)82 cm
Height (Metric) Interior58 cm
Length (English)22.2 in.
Length (Metric)56.5 cm
Temperature Stability±0.3°C
TimerWeekly / Real time / Hour
Wattage3100 w
Width (English) Interior17.2 in.
Width (Metric) Interior43.8 cm
Temperature (Metric)50°C to 330°C
Voltage230 V
Capacity (Metric)99 L
Dimensions (D x W x H) Interior15.3 x 17.2 x 22.8 in. (38.9 x 43.8 x 58 cm)
TypeGravity Convection
Electrical Requirements230 V 50/60 Hz
MaterialStainless Steel
Plug TypeCountry-specific
Width (English)25.2 in.
No. of Shelves2 supplied/16 max.
Width (Metric)64 cm
Unit SizeEach
51028118Full specifications
Each61 LGravity Convection120 V 60 HzCoated Steel
2,891.00
Your Price:
***
Amperage14.4 A
Controller TypeProgrammable Microprocessor control with vacuum fluorescent display
Depth (English) Interior15.3 in.
Depth (Metric) Interior38.9 cm
Dimensions (L x W x H)22.2 x 20.9 x 28.3 in. (56.5 x 53 x 72 cm)
Frequency60 Hz
Height (English)28.3 in.
Height (English) Interior18.9 in.
Height (Metric)72 cm
Height (Metric) Interior48 cm
Length (English)22.2 in.
Length (Metric)56.5 cm
Temperature Stability±0.3°C
TimerWeekly / Real time / Hour
Wattage1730 w
Width (English) Interior12.9 in.
Width (Metric) Interior32.8 cm
Temperature (Metric)50°C to 330°C
Voltage120 V
Capacity (Metric)61 L
Dimensions (D x W x H) Interior15.3 x 12.9 x 18.9 in. (38.9 x 32.8 x 48 cm)
TypeGravity Convection
Electrical Requirements120 V 60 Hz
MaterialCoated Steel
Plug TypeNema 5-15
Width (English)20.9 in.
No. of Shelves2 supplied/39 max.
Width (Metric)53 cm
Unit SizeEach
51028145Full specifications
Each61 LGravity Convection230 V 50/60 HzCoated SteelRequest A Quote***-
Amperage7.9 A
Controller TypeProgrammable Microprocessor control with vacuum fluorescent display
Depth (English) Interior15.3 in.
Depth (Metric) Interior38.9 cm
Dimensions (L x W x H)22.2 x 20.9 x 28.3 in. (56.5 x 53 x 72 cm)
Frequency50/60 Hz
Height (English)28.3 in.
Height (English) Interior18.9 in.
Height (Metric)72 cm
Height (Metric) Interior48 cm
Length (English)22.2 in.
Length (Metric)56.5 cm
Temperature Stability±0.3°C
TimerWeekly / Real time / Hour
Wattage1810 w
Width (English) Interior12.9 in.
Width (Metric) Interior32.8 cm
Temperature (Metric)50°C to 330°C
Voltage230 V
Capacity (Metric)61 L
Dimensions (D x W x H) Interior15.3 x 12.9 x 18.9 in. (38.9 x 32.8 x 48 cm)
TypeGravity Convection
Electrical Requirements230 V 50/60 Hz
MaterialCoated Steel
Plug TypeCountry-specific
Width (English)20.9 in.
No. of Shelves2 supplied/13 max.
Width (Metric)53 cm
Unit SizeEach
51028119Full specifications
Each99 LGravity Convection208/240 V 60 HzCoated Steel
3,131.00
Your Price:
***
Amperage14 A
Controller TypeProgrammable Microprocessor control with vacuum fluorescent display
Depth (English) Interior15.3 in.
Depth (Metric) Interior38.9 cm
Dimensions (L x W x H)22.2 x 25.2 x 32.3 in. (56.5 x 64 x 82 cm)
Frequency60 Hz
Height (English)32.3 in.
Height (English) Interior22.8 in.
Height (Metric)82 cm
Height (Metric) Interior58 cm
Length (English)22.2 in.
Length (Metric)56.5 cm
Temperature Stability±0.3°C
TimerWeekly / Real time / Hour
Wattage3360 w
Width (English) Interior17.2 in.
Width (Metric) Interior43.8 cm
Temperature (Metric)50°C to 330°C
Voltage208/240 V
Capacity (Metric)99 L
Dimensions (D x W x H) Interior15.3 x 17.2 x 22.8 in. (38.9 x 43.8 x 58 cm)
TypeGravity Convection
Electrical Requirements208/240 V 60 Hz
MaterialCoated Steel
Plug TypeNema 6-20
Width (English)25.2 in.
No. of Shelves2 supplied/16 max.
Width (Metric)64 cm
Unit SizeEach
51028146Full specifications
Each99 LGravity Convection230 V 50/60 HzCoated SteelRequest A Quote***-
Amperage13.5 A
Controller TypeProgrammable Microprocessor control with vacuum fluorescent display
Depth (English) Interior15.3 in.
Depth (Metric) Interior38.9 cm
Dimensions (L x W x H)22.2 x 25.2 x 32.3 in. (56.5 x 64 x 82 cm)
Frequency50/60 Hz
Height (English)32.3 in.
Height (English) Interior22.8 in.
Height (Metric)82 cm
Height (Metric) Interior58 cm
Length (English)22.2 in.
Length (Metric)56.5 cm
Temperature Stability±0.3°C
TimerWeekly / Real time / Hour
Wattage3100 w
Width (English) Interior17.2 in.
Width (Metric) Interior43.8 cm
Temperature (Metric)50°C to 330°C
Voltage230 V
Capacity (Metric)99 L
Dimensions (D x W x H) Interior15.3 x 17.2 x 22.8 in. (38.9 x 43.8 x 58 cm)
TypeGravity Convection
Electrical Requirements230 V 50/60 Hz
MaterialCoated Steel
Plug TypeCountry-specific
Width (English)25.2 in.
No. of Shelves2 supplied/16 max.
Width (Metric)64 cm
Unit SizeEach
51028120Full specifications
Each168 LGravity Convection208/240 V 60 HzCoated SteelRequest A Quote***-
Amperage14 A
Controller TypeProgrammable Microprocessor control with vacuum fluorescent display
Depth (English) Interior22.2 in.
Depth (Metric) Interior56.4 cm
Dimensions (L x W x H)29.1 x 25.2 x 36.2 in. (73.8 x 64 x 92 cm)
Frequency60 Hz
Height (English)36.2 in.
Height (English) Interior26.8 in.
Height (Metric)92 cm
Height (Metric) Interior68 cm
Length (English)29.1 in.
Length (Metric)73.8 cm
Temperature Stability±0.3°C
TimerWeekly / Real time / Hour
Wattage3360 w
Width (English) Interior17.2 in.
Width (Metric) Interior43.8 cm
Temperature (Metric)50°C to 330°C
Voltage208/240 V
Capacity (Metric)168 L
Dimensions (D x W x H) Interior22.2 x 17.2 x 26.8 in. (56.4 x 43.8 x 68 cm)
TypeGravity Convection
Electrical Requirements208/240 V 60 Hz
MaterialCoated Steel
Plug TypeNema 6-20
Width (English)25.2 in.
No. of Shelves2 supplied/19 max.
Width (Metric)64 cm
Unit SizeEach
51028147Full specifications
Each168 LGravity Convection230 V 50/60 HzCoated SteelRequest A Quote***-
Amperage13.5 A
Controller TypeProgrammable Microprocessor control with vacuum fluorescent display
Depth (English) Interior22.2 in.
Depth (Metric) Interior56.4 cm
Dimensions (L x W x H)29.1 x 25.2 x 36.2 in. (73.8 x 64 x 92 cm)
Frequency50/60 Hz
Height (English)36.2 in.
Height (English) Interior26.8 in.
Height (Metric)92 cm
Height (Metric) Interior68 cm
Length (English)29.1 in.
Length (Metric)73.8 cm
Temperature Stability±0.3°C
TimerWeekly / Real time / Hour
Wattage3100 w
Width (English) Interior17.2 in.
Width (Metric) Interior43.8 cm
Temperature (Metric)50°C to 330°C
Voltage230 V
Capacity (Metric)168 L
Dimensions (D x W x H) Interior22.2 x 17.2 x 26.8 in. (56.4 x 43.8 x 68 cm)
TypeGravity Convection
Electrical Requirements230 V 50/60 Hz
MaterialCoated Steel
Plug TypeCountry-specific
Width (English)25.2 in.
No. of Shelves2 supplied/19 max.
Width (Metric)64 cm
Unit SizeEach
51028536Full specifications
Each61 LGravity Convection120 V 60 HzStainless Steel
2,874.00
Your Price:
***
Amperage14.4 A
Controller TypeProgrammable Microprocessor control with vacuum fluorescent display
Depth (English) Interior15.3 in.
Depth (Metric) Interior38.9 cm
Dimensions (L x W x H)22.2 x 20.9 x 28.3 in. (56.5 x 53 x 72 cm)
Frequency60 Hz
Height (English)28.3 in.
Height (English) Interior18.9 in.
Height (Metric)72 cm
Height (Metric) Interior48 cm
Length (English)22.2 in.
Length (Metric)56.5 cm
Temperature Stability±0.3°C
TimerWeekly / Real time / Hour
Wattage1730 w
Width (English) Interior12.9 in.
Width (Metric) Interior32.8 cm
Temperature (Metric)50°C to 330°C
Voltage120 V
Capacity (Metric)61 L
Dimensions (D x W x H) Interior15.3 x 12.9 x 18.9 in. (38.9 x 32.8 x 48 cm)
TypeGravity Convection
Electrical Requirements120 V 60 Hz
MaterialStainless Steel
Plug TypeNema 5-15
Width (English)20.9 in.
No. of Shelves2 supplied/13 max.
Width (Metric)53 cm
Unit SizeEach
51028544Full specifications
Each61 LGravity Convection230 V 50/60 HzStainless SteelRequest A Quote***-
Amperage6.1 A
Controller TypeProgrammable Microprocessor control with vacuum fluorescent display
Depth (English) Interior15.3 in.
Depth (Metric) Interior38.9 cm
Dimensions (L x W x H)22.2 x 20.9 x 28.3 in. (56.5 x 53 x 72 cm)
Frequency50/60 Hz
Height (English)28.3 in.
Height (English) Interior18.9 in.
Height (Metric)72 cm
Height (Metric) Interior48 cm
Length (English)22.2 in.
Length (Metric)56.5 cm
Temperature Stability±0.3°C
TimerWeekly / Real time / Hour
Wattage1810 w
Width (English) Interior12.9 in.
Width (Metric) Interior32.8 cm
Temperature (Metric)50°C to 330°C
Voltage230 V
Capacity (Metric)61 L
Dimensions (D x W x H) Interior15.3 x 12.9 x 18.9 in. (38.9 x 32.8 x 48 cm)
TypeGravity Convection
Electrical Requirements230 V 50/60 Hz
MaterialStainless Steel
Plug TypeCountry-specific
Width (English)20.9 in.
No. of Shelves2 supplied/13 max.
Width (Metric)53 cm
Unit SizeEach
51028537Full specifications
Each99 LGravity Convection208/240 V 60 HzStainless SteelRequest A Quote***-
Amperage14 A
Controller TypeProgrammable Microprocessor control with vacuum fluorescent display
Depth (English) Interior15.3 in.
Depth (Metric) Interior38.9 cm
Dimensions (L x W x H)22.2 x 25.2 x 32.3 in. (56.5 x 64 x 82 cm)
Frequency60 Hz
Height (English)32.3 in.
Height (English) Interior22.8 in.
Height (Metric)82 cm
Height (Metric) Interior58 cm
Length (English)22.2 in.
Length (Metric)56.5 cm
Temperature Stability±0.3°C
TimerWeekly / Real time / Hour
Wattage3360 w
Width (English) Interior17.2 in.
Width (Metric) Interior43.8 cm
Temperature (Metric)50°C to 330°C
Voltage208/240 V
Capacity (Metric)99 L
Dimensions (D x W x H) Interior15.3 x 17.2 x 22.8 in. (38.9 x 43.8 x 58 cm)
TypeGravity Convection
Electrical Requirements208/240 V 60 Hz
MaterialStainless Steel
Plug TypeNema 6-20
Width (English)25.2 in.
No. of Shelves2 supplied/16 max.
Width (Metric)64 cm
Unit SizeEach
Showing 10 of 23Show more
Features:
  • Broad temperatures range — up to 330°C
  • Advanced digital timer for daily or weekly on/off cycles
  • Stainless-steel interior is easy to clean and corrosion resistant (steel quality AISI 304)
  • Programmable controller for temperature ramps and dwells
  • Up to 10 programs can be saved, each with 10 discrete steps
  • Electronically controlled fan speed and damper position can be incorporated
  • Programs can be repeated automatically
  • Access port allows the introduction of sensors for independent data monitoring
  • Simple calibration routine ensures temperature accuracy over time
  • Boost function enables rapid heating — no need to operate the oven 24/7
Ordering Information:
Availability may vary by country.
Warranty:
2-year warranty
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.