Cytomat™ 10 C425 Series Automated Incubators
Actual product may vary
Thermo Scientific™

Cytomat™ 10 C425 Series Automated Incubators

Temperature Range: 4°C to 25°C with External Chiller; Relative Humidity: <90% at 25°C
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVoltage
51031161100 V
51031162120 V
51031164230 V
Catalog number 51031161
Price (USD)
-
Voltage:
100 V

Designed to fit into the busiest of cell biology or screening laboratories, the Thermo Scientific™ Cytomat™ 10 C425 Series Automated Incubator is a low-temperature-range incubator and storage unit. It is ideal for high capacity incubation (210 SBS microplates needing a temperature range of up to 25°C combined with 4°C storage. The Cytomat 10 C425 incubator seamlessly integrates with existing automated assay hardware, and provides mean access time below 10 seconds, controlled humidity, and ContraCon thermal decontamination routine.

10 Stacker to Match Your Capacity

  • 280 pcs. 1536-well plates
  • 210 pcs. 96-well or 384-well plates
  • 150 pcs. transwell or insert plates
  • 100 pcs. deep-well

Mix and match different pitch stackers. Additional stacker types are available upon request.

Effortless Humidity Control

  • HydraSmart Technology provides ultrasonic vapor technology with no turbulence and heat introduction into system
  • Sterilized water supply by external accessible micro-particle filter
  • Closed loop water delivery system, with simple external tank
  • Water level monitoring with refill warning

Fast

  • Below 10sec. access time
  • Application speed settings: slow, medium, fast
  • Individual microplate speed settings

Contamination Prevention

  • ContraCon automated decontamination routine with true walk-away time
  • Easy-to-clean, smooth stainless steel surface
  • Solid copper inner chamber (option)
  • H2O2 flange (option)

Flexible Stacker Design

  • Compatible with existing Cytomat stacker
  • Standard or customized stacker pitch sizes
  • Mix and match different pitch stackers
  • Autoclaveable stainless steel
  • Ventilation slots allow accurate temperature distribution

Convenient, Easy to Use

  • 5-inch touch screen with LED backlight as standard
  • Various gate / transfer positions

Easy to Integrate

  • Intuitive software commands
  • Flat gate, no gate cover
  • Designed with multiple transfer stations

More options

  • O2 control
  • Solid copper inner chamber
  • H2O2 flange
  • Internal and direct barcode reader

Certification

  • CE
  • RoHS III
  • Manufactured in ISO 9001 and ISO 13485 accredited facility
Specifications
Access TimeBelow 10 sec.
Certifications/ComplianceCE marked
Depth (English)32.3 in.
Depth (Metric)822 mm
Electrical Requirements100 V, 50 Hz
FeaturesIncubation and storage unit with temperature range below with external chiller, humidity up to 90% r.H. at 25°C
Height (English)36.0 in.
Height (Metric)915 mm
Holds280pcs. 1536-well plates
210pcs. 96-well or 384-well plates
150pcs. transwell or insert plates
100pcs. deep-well
TypeAutomated Incubator
Width (English)31.5 in.
Width (Metric)800 mm
Chamber MaterialStainless Steel
MaterialStainless Steel
Relative Humidity<90%
Temperature Range (Metric)4°C to 25°C (with External Chiller)
Voltage100 V
Unit SizeEach