Benchtop Coolers
Benchtop Coolers
Benchtop Coolers
Benchtop Coolers
Benchtop Coolers
Benchtop Coolers
Thermo Scientific™

Benchtop Coolers

Protect enzymes, cells, reagents and solutions by maintaining freezer temperatures while on the laboratory bench using benchtop coolers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTemperature RangeArrayHolds
5115-0012-20°C3 x 412 Tubes (0.2 to 0.5 mL)
355501-20°C4 x 520 Tubes (0.2 to 0.5 mL)
5115-0032-20°C4 x 832 Tubes (0.2 to 0.5 mL)
5116-13000°C3 x 412 Tubes (12 to 13 mm dia.)
5116-16000°C3 x 412 Tubes (16 to 17 mm dia.)
DS5116-00120°C3 x 412 Cryogenic Vials, Microcentrifuge Tubes (0.2 to 0.5 mL)
DS5116-00320°C4 x 832 Cryogenic Vials, Microcentrifuge Tubes (0.2 to 0.5 mL)
DS5114-0012-20°C3 x 412 Cryogenic Vials (0.2 to 1.5 mL)
Catalog number 5115-0012
Price (KRW)
176,000
Online offer
Ends: 31-Dec-2026
207,000
Save 31,000 (15%)
Each
Temperature Range:
-20°C
Array:
3 x 4
Holds:
12 Tubes (0.2 to 0.5 mL)
Price (KRW)
176,000
Online offer
Ends: 31-Dec-2026
207,000
Save 31,000 (15%)
Each

Protect enzymes, cells, reagents and solutions by maintaining freezer temperatures while on the laboratory bench using Thermo Scientific™ Benchtop Coolers.

  • Maintain freezer temperature during frost-free cycles, power failures and accidental door openings
  • Holds 12, 20 or 32 tubes
  • Inserts are included for 1.5 mL tubes to have the compatibility to hold 0.2-0.5 mL microcentrifuge tubes
  • Printed grid on one-way keyed lid for proper orientation and sample identification
  • Molded of impact-resistant polycarbonate with locking handle for long-term use
  • Stackable to save space
Specifications
Array3 x 4
Description3x4 compartments, -20 deg.
DesignNon-filled/clear lid
Dimensions (L x W x H)5.31 x 4.25 x 4.94 in. (151 x 108 x 125 mm)
Height (English)4.94 in.
Height (Metric)125 mm
Holds Temp., Hours<-15°C, 1
Length (English)5.31 in.
Length (Metric)151 mm
Width (English)4.25 in.
Width (Metric)108 mm
Capacity (Metric)0.2 to 0.5 mL
ColorBlue, Clear
Holds12 Tubes (0.2 to 0.5 mL)
MaterialPolycarbonate
Product TypeBenchtop Cooler
Temperature Range-20°C
Unit SizeEach