Product Image
Thermo Scientific™
Thermo Scientific™ QSP™ Amber Screw Cap Micro Tubes
Thermo Scientific™ QSP™ Screw Cap Micro Tubes robust design and material selection makes them great with biologics, sample preparation, storage and transportation.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Bottom Shape
514-A-Q1.5 mLConical
516-A-Q0.5 mLConical (Self-standing)
519-GRD-A-Q2.0 mLConical (Self-standing)
521-A-Q2.0 mLConical
528-A-Q1.5 mLConical (Self-standing)
Catalog number 514-A-Q
Price (BRL)
-
Capacity (Metric):
1.5 mL
Bottom Shape:
Conical
Thermo Scientific™ QSP™ Amber Screw Cap Micro Tubes, 0.5ml, knurled, conical, Amber, tube only no cap. Packaged in bulk bags and non-sterile clean claims are; RNase/DNase, DNA, pyrogen and endotoxin free.
Their robust design and material selection are well suited for work with biologics and offer rich features, including ultra-clear externally threaded tubes and UV protective amber tubes.
  • Screw Cap micro tube volume range of 0.5, 1.5 and 2.0ml and are ideal for sample preparation, storage and transportation
  • Conical bottom or self standing offered with knurls in ultra clear natural or UV protective amber color
  • Tether caps and standard non tether caps are available in a rainbow of color options
Specifications
Clean ClaimsRNase/DNase, DNA, pyrogen and endotoxin free
AutoclavableAutoclavable
Bottom ShapeConical
Capacity (Metric)1.5 mL
ColorAmber
GraduatedYes
MaterialPolypropylene
SterilityNon-sterile
Unit SizeCase of 10