Product Image
Thermo Scientific™
O2 Control
For Use With: Heracell™ VIOS 160i and Forma™ Steri-Cycle™ i160 Incubators
Have Questions?
Catalog NumberOxygen Control
519011371 to 21%
Catalog number 51901137
Price (MXN)
-
Oxygen Control:
1 to 21%
Upgrade your incubators to take advantage of O2 control.
Specifications
Description1 to 21% O2 control
For Use With (Equipment)Heracell™ VIOS 160i and Forma™ Steri-Cycle™ i160 Incubators
LightingLED
Oxygen Control1 to 21%
TypeO2 Control
Unit SizeEach