Opti-MEM™ I Reduced Serum Medium, GlutaMAX™ Supplement
Opti-MEM™ I Reduced Serum Medium, GlutaMAX™ Supplement
Gibco™

Opti-MEM™ I Reduced Serum Medium, GlutaMAX™ Supplement

Opti-MEM™ I Reduced-Serum Medium is an improved Minimal Essential Medium (MEM) that allows for a reduction of Fetal Bovine SerumRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
5198509110 x 500 mL
51985034500 mL
Catalog number 51985091
Price (USD)
658.65
Online Exclusive
716.00
Save 57.35 (8%)
Each
Add to cart
Quantity:
10 x 500 mL
Customize this product
Price (USD)
658.65
Online Exclusive
716.00
Save 57.35 (8%)
Each
Add to cart

Opti-MEM™ I Reduced-Serum Medium is an improved Minimal Essential Medium (MEM) that allows for a reduction of Fetal Bovine Serum supplementation by at least 50% with no change in growth rate or morphology. Opti-MEM™ I medium is also recommended for use with cationic lipid transfection reagents, such as Lipofectamine™ reagent. Opti-MEM™ I medium can be used with a variety of suspension and adherent mammalian cells, including Sp2, AE-1, CHO, BHK-21, HEK, and primary fibroblasts. We offer a variety of Opti-MEM™ I modifications for a range of cell culture applications.


This Opti-MEM™ I is modified as follows:
With
• GlutaMAX™
• Phenol Red


The complete formulation is confidential. For more information, please contact Technical Services.

Using Opti-MEM™ I Medium
Opti-MEM™ I Reduced-Serum Medium is a unique medium that contains insulin, transferrin, hypoxanthine, thymidine, and trace elements. These additional components allow for a reduction in serum supplementation by at least 50%. Opti-MEM™ I medium uses a sodium bicarbonate buffer system (2.4 g/L), and therefore requires a 5–10% CO2 environment to maintain physiological pH.

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
Cell LineSp2, AE-1, CHO, BHK-21, and HEK
Cell TypePrimary Fibroblasts
Concentration1X
Manufacturing QualitycGMP-compliant under the ISO 13485 standard
Product LineGibco, GlutaMAX, Opti-MEM
Product TypeOpti-MEM I
Quantity10 x 500 mL
Shelf Life18 Months From Date of Manufacture
Shipping ConditionRoom Temperature
ClassificationAnimal Origin
FormLiquid
SterilitySterile-filtered
With AdditivesPhenol Red
Unit SizeEach
Contents & Storage
Storage conditions: 2-8° C. Protect from light
Shipping conditions: Ambient
Shelf life: 18 months from date of manufacture

Frequently asked questions (FAQs)

Is there any other difference between Opti-MEM I Reduced Serum Medium and Opti-MEM I Reduced Serum Medium, no phenol red besides the presence/absence of phenol red?

The formulation of Opti-MEM I Reduced Serum Medium and that of Opti-MEM I Reduced Serum Medium, no phenol red is the same except for the presence of phenol red in the former medium.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Where can I find the osmolality for my lot of Opti-MEM I Reduced Serum Medium?

We do provide osmolality information on the certificate of analysis. All lots of Opti-MEM I Reduced Serum Medium will meet the osmolality specification of ≥280 to ≤320 mOsm/kg.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Can I aliquot and freeze Opti-MEM I Reduced Serum Medium to conserve it longer? If so, for how long can it be stored?

Sorry, we don't have data to show that frozen Opti-MEM I Reduced Serum Medium has a longer shelf life. Actually, we recommend storing this medium at 2-8 degrees C in the dark. We do not recommend freezing the medium because it may precipitate.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

I noticed that Opti-MEM I Reduced Serum Medium does not contain any serum. Why do you then refer to it as "Reduced Serum Medium"?

Opti-MEM I Reduced Serum Medium is a serum-free medium. The reason why it is called “reduced-serum medium” is because you only need to supplement this medium with 1-5% FBS instead of the standard 10% FBS.

Opti-MEM I Reduced Serum Medium can be used as is (without serum) for transfection of cells. However, for long-term cell culture, you need to add serum to the medium to keep cells healthy.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

How long can I keep my media after supplementing with serum?

Generally speaking, media can be used for up to three weeks after supplementation with serum. There are no formal studies to support this, but it is the rule of thumb used by our scientists.

Find additional tips, troubleshooting help, and resources within our Mammalian Cell Culture Basics Support Center.