Search
Search
| Catalog Number | Height (Metric) | Size | For Use With | Unit Size |
|---|---|---|---|---|
| 5242-0040 | 51.4 cm | A | Pipet, 8 in. | Case of 6 |
| 5242-0040PK also known as 5242-0040 | 51.4 cm | A | Pipet, 8 in. | Each |
| 5242-0050 | 68.6 cm | B | Pipet, 16 in. | Case of 6 |
| 5242-0050PK also known as 5242-0050 | 68.6 cm | B | Pipet, 16 in. | Each |
| 5242-0060 | 91.4 cm | C | Pipet, 24 in. | Case of 2 |
| 5242-0060PK | 91.4 cm | C | Pipet, 24 in. | Each |
Soak or sterilize glass pipettes using Thermo Scientific™ Nalgene™ Pipette Jars. Fill jars with preferred cleaning solution then place pipettes in the corresponding Nalgene pipette basket (Part No. DS5241) for easy submersion in and retrieval from the jar. The lid slows cleaning solution evaporation and keeps contents free from debris. The jars feature a flanged base for added security.
Recycling information:
Please recycle the corrugate cartons and any LDPE bags your Nalgene products are delivered in.